Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
251-385-9
MDL number:
Pricing and availability is not currently available.
Product Name
Etomidate, >98% (HPLC), powder
Quality Level
assay
>98% (HPLC)
form
powder
color
white
solubility
DMSO: >10 mg/mL
originator
Johnson & Johnson
storage temp.
2-8°C
SMILES string
CCOC(=O)c1cncn1[C@H](C)c2ccccc2
InChI
1S/C14H16N2O2/c1-3-18-14(17)13-9-15-10-16(13)11(2)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/t11-/m1/s1
InChI key
NPUKDXXFDDZOKR-LLVKDONJSA-N
Gene Information
General description
Etomidate is an imidazole derivative, which functions as an nonbarbiturate hypnotic agent. It is used in speech and memory test. Etomidate regulates hemodynamic stability by preserving sympathetic outflow and autonomic reflexes. It inhibits adrenal steroidogenesis.
Biochem/physiol Actions
Etomidate is a general anesthetic; potentiates GABAA transmission
Etomidate is a general anesthetic; potentiates GABAA transmission. The possible neuroprotective effect of etomidate against streptozotocin-induced (STZ-induced) hyperglycaemia were investigated in the rat brain and spinal cord. Etomidate treatment demonstrated neuroprotective effect on the neuronal tissue against the diabetic oxidative damage
Features and Benefits
This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
1 of 1
This Item | |||
|---|---|---|---|
| assay >98% (HPLC) | assay - | assay - | assay ≥98% (HPLC) |
| form powder | form - | form - | form oil |
| Quality Level 100 | Quality Level - | Quality Level - | Quality Level - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: >10 mg/mL | solubility - | solubility - | solubility - |
| originator Johnson & Johnson | originator - | originator - | originator - |
Still not finding the right product?
Explore all of our products under Etomidate
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Laura C McPhee et al.
Critical care medicine, 41(3), 774-783 (2013-01-16)
Retrospective analyses of several trials suggest etomidate may be unsafe for intubation in patients with sepsis. We evaluated the association of etomidate and mortality in a large cohort of septic patients to determine if single-dose etomidate was associated with increased
Etomidate speech and memory test (eSAM) A new drug and improved intracarotid procedure
Jones-Gotman M, et al.
Neurology, 65(11), 1723-1729 (2005)
Beatrice de la Grandville et al.
European journal of anaesthesiology, 29(11), 511-514 (2012-09-12)
Etomidate is an imidazole-derived hypnotic agent preferentially used for rapid sequence induction of anaesthesia because of its favourable haemodynamic profile. However, 11β-hydroxylase inhibition causes adrenal insufficiency with potentially fatal consequences in specific populations. We review the arguments against the liberal
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E6530-10MG | 04061832090719 |
| E6530-50MG | 04061832090726 |



