Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-773-7
MDL number:
Specific activity:
≥15 units/mg solid
Pricing and availability is not currently available.
form
lyophilized powder
Quality Level
specific activity
≥15 units/mg solid
mol wt
168 kDa
application(s)
diagnostic assay manufacturing
storage temp.
−20°C
Application
Esterase from porcine liver has been used in a study to assess the effect of 5-aminolaevulinic acid peptide prodrugs on photosensitization for photodynamic therapy. Esterase from porcine liver has also been used in a study to investigate how site-specific atherogenic gene expression correlates with subsequent variable lesion development in coronary and peripheral vasculature.
Porcine liver esterase is used to catalyze the hydrolysis of pentaacetyl catechin and epicatechin for use in pharmaceutical and industrial applications.
Pig liver esterase is commonly used for kinetic resolutions and assymetric synthesis in organic chemistry.
Pig liver esterase is commonly used for kinetic resolutions and assymetric synthesis in organic chemistry.
The enzyme from Sigma has been immobilised in hollow fibre ultrafiltration membrane and used for the asymmetric hydrolysis of a meso-diester. Esterase from Sigma has been used to investigate its effect on the release of methacrylic acid (MAA) and 2-hydroxyethyl methacrylate (HEMA) from adhesives formulated under conditions simulating wet bonding.[1] It has been used to examine the ability of carboxylesterase activity to remove permethrin- and bifenthrin-associated toxicity to Ceriodaphnia dubia and Hyalella azteca in a variety of matrices.[2]
Biochem/physiol Actions
Esterase acts on water-soluble carboxyl esters containing short chain fatty acids. Its functionality is attributed to the catalytic triad of Ser, His and Asp/Glu.
Pig liver esterase catalyzes enantioselective conversion of an ester to a carboxylic acid. The molecular weight is found to be 168 kDa. It is a serine enzyme with two active sites on each molecule, which dissociates into active half-molecules in the presence of dilute acid or concentrated salts.[3]
Other Notes
One unit will hydrolyze 1.0 μmole of ethyl butyrate to butyric acid and ethanol per min at pH 8.0 at 25 °C.
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ≥15 units/mg solid | specific activity ≥30 units/mg protein | specific activity ≥30.0 U/g | specific activity ≥0.2 U/mg |
| application(s) diagnostic assay manufacturing | application(s) - | application(s) - | application(s) - |
| form lyophilized powder | form lyophilized powder | form crystalline, crystals, powder or flakes | form lyophilized |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| mol wt 168 kDa | mol wt 60 kDa | mol wt - | mol wt - |
| Quality Level 300 | Quality Level 200 | Quality Level 100 | Quality Level - |
Still not finding the right product?
Explore all of our products under Esterase from porcine liver
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Enzyme-responsive snap-top covered silica nanocontainers.
Kaushik Patel et al.
Journal of the American Chemical Society, 130(8), 2382-2383 (2008-02-01)
Elisabet L Kostoryz et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 88(2), 394-401 (2008-04-09)
Dentin adhesives may undergo phase separation when bonding to wet demineralized dentin. We hypothesized that adhesives exhibiting phase separation will experience enhanced biodegradation of methacrylate ester groups. The objective of this project was to study the effect of enzyme-exposure on
Anatoli Tchigvintsev et al.
Applied microbiology and biotechnology, 99(5), 2165-2178 (2014-09-10)
Most of the Earth's biosphere is cold and is populated by cold-adapted microorganisms. To explore the natural enzyme diversity of these environments and identify new carboxylesterases, we have screened three marine metagenome gene libraries for esterase activity. The screens identified
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E3019-3.5KU | 04061833603093 |
| E3019-20KU | 04061833603086 |
| E3019-100KU | 04061833603079 |




