Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Esterase from porcine liver
Synonym(s):
Carboxyl esterase, Carboxylic-ester hydrolase, PLE
CAS Number:
9016-18-6
EC Number:
232-773-7
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Physical Form
Tissue
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
E3019
Esterase from porcine liver,
lyophilized powder, ≥15 units/mg solid
Expand
View Pricing
E2884
Esterase from porcine liver,
ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Expand
View Pricing
46058
Esterase from porcine liver,
lyophilized, powder, slightly beige, ≥50 U/mg
Expand
View Pricing