Skip to Content
Merck

Skip To

D5899

2-Deoxy-D-ribose

BioReagent, suitable for cell culture

Synonym(s):

2-Deoxy-D-arabinose, 2-Deoxy-D-erythropentose, Thyminose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
UNSPSC Code:
12352201
NACRES:
NA.75
PubChem Substance ID:
EC Number:
208-573-0
Beilstein/REAXYS Number:
1721978
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

product line

BioReagent

assay

≥99% (GC)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

89-90 °C (lit.)

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)CC=O

InChI

1S/C5H10O4/c6-2-1-4(8)5(9)3-7/h2,4-5,7-9H,1,3H2/t4-,5+/m0/s1

InChI key

ASJSAQIRZKANQN-CRCLSJGQSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
31170D3179121649
technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

gas chromatography (GC): suitable

technique(s)

-

biological source

synthetic (organic)

biological source

-

biological source

synthetic (organic)

biological source

-

assay

≥99% (GC)

assay

≥99.0% (TLC)

assay

≥98% (GC)

assay

97% (GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to light yellow

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

-

form

powder

form

solid

form

powder

form

powder


Still not finding the right product?

Explore all of our products under 2-Deoxy-D-ribose


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
D5899-1G04061833586990
D5899-10MG04061833586983

Questions

Reviews

No rating value

Active Filters