Skip to Content
Merck
All Photos(4)

Key Documents

D3179

Sigma-Aldrich

2-Deoxy-D-glucose

≥98% (GC), BioXtra

Synonym(s):

2-Deoxy-D-arabinohexose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
Beilstein:
1723331
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

product line

BioXtra

Assay

≥98% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

impurities

≤0.001% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

color

white

mp

146-147 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.003%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)CC=O

InChI

1S/C6H12O5/c7-2-1-4(9)6(11)5(10)3-8/h2,4-6,8-11H,1,3H2/t4-,5-,6+/m1/s1

InChI key

VRYALKFFQXWPIH-PBXRRBTRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Deoxy-D-glucose was used in the development of anti-cancer strategies that involve radio- and chemosensitization and oxidative stress. It was used in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR).

Biochem/physiol Actions

2-Deoxy-D-Glucose (2-Deoxyglucose) is a glucose analog that inhibits glycolysis via its action on hexokinase, the rate limiting step of glycolysis. It is phosphorylated by hexokinase to 2-DG-P which can not be further metabolized by phosphoglucose isomerase. This leads to the accumulation of 2-DG-P in the cell and the depletion in cellular ATP. In vitro, 2-Deoxyglucose has been shown to induce autophagy, increases ROS production, and activate AMPK.
2-Deoxy-D-Glucose (2-Deoxyglucose) is a glucose analog that inhibits glycolysis via its actions on hexokinase, the rate limiting step of glycolysis. It is phosphorylated by hexokinase to 2-DG-P which can not be further metabolized by phosphoglucose isomerase. This leads to the accumulation of 2-DG-P in the cell and the depletion in cellular ATP. In vitro, 2-Deoxyglucose has been shown to induce autophagy, increase ROS production, and activate AMPK.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig Beall et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 302(2), R215-R223 (2011-11-11)
Despite significant technological and pharmacological advancements, insulin replacement therapy fails to adequately replicate β-cell function, and so glucose control in type 1 diabetes mellitus (T1D) is frequently erratic, leading to periods of hypoglycemia. Moreover, the counterregulatory response (CRR) to falling
B S Dwarakanath
Journal of cancer research and therapeutics, 5 Suppl 1, S27-S31 (2009-12-17)
The glucose analog 2-deoxy-D-glucose (2-DG), an inhibitor of glucose transport and glycolytic ATP production, is the most widely investigated metabolic inhibitor for targeting glucose metabolism. Besides depleting energy in cells, 2-DG has also been found to alter N-linked glycosylation leading
Abdullah Farooque et al.
Journal of cancer research and therapeutics, 5 Suppl 1, S32-S35 (2009-12-17)
Normal tissue toxicity is one of the major limiting factors in cancer therapy. Damage to normal tissues and critical organs restricts the use of higher therapeutic doses thereby compromising the efficacy. The glucose analog 2-deoxy-D-glucose (2-DG), an inhibitor of glycolytic
Madhusudhanan Sukumar et al.
The Journal of clinical investigation, 123(10), 4479-4488 (2013-10-05)
Naive CD8+ T cells rely upon oxidation of fatty acids as a primary source of energy. After antigen encounter, T cells shift to a glycolytic metabolism to sustain effector function. It is unclear, however, whether changes in glucose metabolism ultimately
Rosemarie Ungricht et al.
The Journal of cell biology, 209(5), 687-703 (2015-06-10)
Newly synthesized membrane proteins are constantly sorted from the endoplasmic reticulum (ER) to various membranous compartments. How proteins specifically enrich at the inner nuclear membrane (INM) is not well understood. We have established a visual in vitro assay to measure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service