Skip to Content
Merck

Skip To

D1556

OptiPrep Density Gradient Medium

used for cell and subcellular organelle isolation

OptiPrep™ Density Gradient Medium

Synonym(s):

Iodixanol solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.75
PubChem Substance ID:
UNSPSC Code:
12352207
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile

form

liquid

density

1.319-1.321 g/mL

application(s)

hematology
histology

storage temp.

room temp

SMILES string

CC(=O)N(CC(O)CN(C(C)=O)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I)c2c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c2I

InChI

1S/C35H44I6N6O15/c1-13(52)46(30-26(38)20(32(59)42-3-15(54)9-48)24(36)21(27(30)39)33(60)43-4-16(55)10-49)7-19(58)8-47(14(2)53)31-28(40)22(34(61)44-5-17(56)11-50)25(37)23(29(31)41)35(62)45-6-18(57)12-51/h15-19,48-51,54-58H,3-12H2,1-2H3,(H,42,59)(H,43,60)(H,44,61)(H,45,62)

InChI key

NBQNWMBBSKPBAY-UHFFFAOYSA-N

General description

OptiPrep Density Gradient Medium, also known as iodixanol, is a low osmolarity, non-ionic iodinated density gradient medium. It is a sterile and endotoxin-tested solution that comprises 60% (w/v) iodixanol in water, designed for the in vitro isolation of biological particles. It may be used for fractionation and separation of cell and subcellular organelles.[1]

Application

OptiPrep has been used to isolate molecules, cells, and subcellular organelles, including DNA, cytokines, lysosomes, endoplasmic reticulum, peroxisomes, T lymphocytes, and natural killer cells.[1] It has also been used for the following applications:

  • to investigate the efficiency of single-cell encapsulation.[1]
  • the gradient-based isolation of senescent cancer cells.
  • isolation of non-enveloped virion isolation from bronchoalveolar lavage.
  • isolation of lipoproteins and macromolecules
  • purification of virions
  • purification of neuron cells

Features and Benefits

OptiPrep has numerous biocompatible properties that make it ideal for use.
  • It is biologically inert and does not interfere with assays for marker enzymes.[1]
  • It is non-ionic, non-toxic, and has low osmotic pressure.
  • It forms spontaneous gradients that are stabler and easier to prepare than sucrose.

Other Notes

60% (w/v) solution of iodixanol in water (sterile).

Legal Information

OptiPrep is a trademark of Serumwerk Bernburg AG

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I1284T9285A6964
form

liquid

form

liquid

form

liquid

form

liquid

density

1.319-1.321 g/mL

density

-

density

-

density

-

sterility

sterile

sterility

-

sterility

0.2 μm filtered

sterility

sterile-filtered

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

room temp

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C

application(s)

hematology
histology

application(s)

-

application(s)

sample preparation

application(s)

-


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



WU Polyomavirus in Respiratory Epithelial Cells from Lung Transplant Patient with Job Syndrome
Erica A. S, et al.
Emerging Infectious Diseases, 21(1), 103-106 (2015)
Tanner C Fadero et al.
eLife, 6 (2017-09-05)
Iodixanol provides an easy and affordable solution to a problem that has limited resolution and brightness when imaging living samples.
Isolation of senescent cells by iodixanol (OptiPrep) density gradient-based separation
Kristina Kovacovicova, Manlio Vinciguerra
Cell Proliferation, 52 (2019)



Global Trade Item Number

SKUGTIN
D1556-100ML04061837280368
D1556-250ML04061838261700

Questions

1–9 of 9 Questions  
  1. What is the density of OptiPrep: 60% v/v, w/w, or w/v?

    1 answer
    1. The density of OptiPrep is expressed as weight/volume (w/v). The stock concentration from the bottle is 60%.

      Helpful?

  2. What is the osmolarity of OptiPrep?

    1 answer
    1. According to the supplier information, the osmolarity of OptiPrep is stated to be 170 mOsm with a deviation of +/- 15 mOsm.

      Helpful?

  3. What is the molarity of OptiPrep (catalog number D1556)?

    1 answer
    1. The molecular weight of OptiPrep is 1552.2 amu, and its density is 1.320 g/ml. The prepared solution contains 60% OptiPrep. Based on these values, the calculated molarity is 0.706 Molar.

      Helpful?

  4. What is the method used to determine the density mentioned on the Certificate of Analysis (CoA) for D1556 OptiPrep™ Density Gradient Medium?

    1 answer
    1. Based on the provided details, utilizing a densitometer or pycnometer is recommended.

      Helpful?

  5. What is the viscosity of the solution?

    1 answer
    1. This product is not tested for viscosity. This item is a 60% solution of Iodixanol in water. However, Iodixanol properties reported by drugfuture.com state that a 50% solution has a viscosity of 8.6 mPa at 37°C.

      Helpful?

  6. How do I store solutions of Product D1556, OptiPrep™ Density Gradient Medium?

    1 answer
    1. After diluting Product D1556, OptiPrep™ Density Gradient Medium, to the required density, the solution can be stored at 4°C. It should be mixed well before use to make sure it has not settled.

      Helpful?

  7. What is the appearance of Product D1556, OptiPrep?

    1 answer
    1. The solution may be clear to slightly yellow.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  9. Why choose Product D1556, OptiPrep, instead of other density gradients such as Percoll or Histopaque?

    1 answer
    1. Percoll and Histopaque must be removed prior to use through washing.  If Percoll and Histopaque are not removed, these density gradients can be toxic to mammalian cells.  In comparison, OptiPrep has no potential for bioaccumulation and can be considered nontoxic to cells. OptiPrep also has lower viscosity as compared to polysucrose based products such as Histopaque or Ficoll.

      Helpful?

Reviews

Active Filters

  1. Atlanta, GA
    • Reviews 3
    • Votes 95
    3 out of 5 stars.

    Expensive OptiPrep

    It worked as OptiPrep from other companies, but it's over-priced, in my opinion.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for taking the time to leave a review! Our Customer Service department is available to discuss getting the best quality products for your needs at the right price. Please feel free to contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Product Pricing and Availability ticket. We look forward to hearing from you soon!