Skip to Content
Merck
All Photos(3)

Key Documents

T9285

Sigma-Aldrich

Tris-EDTA buffer solution

100X concentrate for Northern and Southern blotting

Synonym(s):

Tris-EDTA wash buffer, TE buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

liquid

pH

7.8-8.2 (1 × in water)

application(s)

sample preparation

foreign activity

DNase, RNase, none detected

storage temp.

room temp

SMILES string

NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2

InChI key

VLEIUWBSEKKKFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris EDTA buffer is commonly used in molecular biology for reconstitution and storage of many precipitated biomolecules, including DNA and RNA. Tris-HCl is a commonly used buffer and EDTA is a metal chelating agent. TE buffer solubilizes nucleic acids while protecting the molecules from degradation. It is used in nucleic acid isolation, which may be done prior to nucleic acid gel electrophoresis and Northern or Southern blot hybridization.

Application

TE buffer has been used for:
  • isolate, resuspend, or store DNA and RNA for PCR or qPCR
  • gel electrophoresis, including Northern and Southern blotting
  • bacterial cells, for AmpC disk test, to measure β-lactamases released from the cells
  • antigen retrieval for immunohistochemistry

Components

TE Buffer contains 1M Tris-HCl (pH approximately 8.0), containing 0.1M EDTA.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Black et al.
Journal of clinical microbiology, 43(7), 3110-3113 (2005-07-08)
Although plasmid-mediated AmpC beta-lactamases were first reported in the late 1980s, many infectious disease personnel remain unaware of their clinical importance. These enzymes are typically produced by isolates of Escherichia coli, Klebsiella spp., Proteus mirabilis, and Salmonella spp. and are
Virtual Fluorescence Color Channels by Selective Photobleaching in Digital PCR Applied to the Quantification of KRAS Point Mutations
Schlenker F et al.
Analytical Chemistry, 93, 10538-10545 (2021)
Sarah Bøje et al.
Immunology and cell biology, 94(2), 185-195 (2015-08-14)
Chlamydia is the most widespread sexually transmitted bacterial disease and a prophylactic vaccine is highly needed. Ideally, this vaccine is required to induce a combined response of Th1 cell-mediated immune (CMI) response in concert with neutralizing antibodies. Using a novel
Marika Bogdani et al.
Scientific reports, 7(1), 17231-17231 (2017-12-10)
Cystic fibrosis (CF)-related diabetes (CFRD) is thought to result from beta-cell injury due in part to pancreas exocrine damage and lipofibrosis. CFRD pancreata exhibit reduced islet density and altered cellular composition. To investigate a possible etiology, we tested the hypothesis
Shiraz Tyebji et al.
Cell reports, 31(4), 107573-107573 (2020-04-30)
Germline epigenetic factors influence transgenerational inheritance of behavioral traits upon changes in experience and environment. Immune activation due to infection can also modulate brain function, but whether this experience can be passed down to offspring remains unknown. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service