Skip to Content
Merck

Skip To

D0819

DMEM - high glucose

StableCell, with stable glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

suitability

suitable for

components

stable glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: no
glucose: high
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

General description

This DMEM-Hi glucose medium differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0822D0697FG0435
form

liquid

form

liquid

form

liquid

form

liquid

suitability

suitable for

suitability

-

suitability

-

suitability

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

components

stable glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: no
glucose: high
NaHCO3: yes

components

glucose: high
HEPES: no
stable glutamine
sodium pyruvate: yes
phenol red: yes
NaHCO3: yes

components

glucose: high
sodium pyruvate: yes
stable glutamine: yes
NaHCO3: yes
phenol red: yes
HEPES: yes

components

phenol red: yes
glucose: high
stable glutamine: yes
sodium pyruvate: no
HEPES: no
NaHCO3: yes

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under StableCell DMEM - high glucose


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hiba Msheik et al.
Placenta, 90, 90-97 (2020-02-15)
The placenta is a transitory organ essential for proper fetal maturation and growth. Trophoblasts, the main cell type of the placenta, differentiate along the villous or extravillous pathways. The ability of villous cytotrophoblasts to undergo an epithelial-to-mesenchymal transition to form
Jaroslav Čapek et al.
Materials science & engineering. C, Materials for biological applications, 58, 900-908 (2015-10-20)
An interest in biodegradable metallic materials has been increasing in the last two decades. Besides magnesium based materials, iron-manganese alloys have been considered as possible candidates for fabrication of biodegradable stents and orthopedic implants. In this study, we prepared a
Lingling Teng et al.
Medical science monitor : international medical journal of experimental and clinical research, 26, e927305-e927305 (2020-10-24)
BACKGROUND Preeclampsia (PE) remains one of the primary causes of maternal morbidity and mortality worldwide. This study was designed to investigate the relevance of long non-coding RNA (lncRNA) nuclear-enriched abundant transcript 1 (NEAT1) and downstream molecules in trophoblast cell proliferation



Global Trade Item Number

SKUGTIN
D0819-500ML04061833558140

Questions

Reviews

No rating value

Active Filters