![MegaCell™ Dulbecco’s Modified Eagle’s Medium liquid, sterile-filtered, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/409/249/587be5dd-1236-43cb-b0a7-e02b95c6764a/640/587be5dd-1236-43cb-b0a7-e02b95c6764a.jpg)
DMEM - high glucose
![MegaCell™ Dulbecco’s Modified Eagle’s Medium liquid, sterile-filtered, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/409/249/587be5dd-1236-43cb-b0a7-e02b95c6764a/640/587be5dd-1236-43cb-b0a7-e02b95c6764a.jpg)
Synonym(s):
DME, DMEM, Dulbecco’s Modified Eagle’s Medium - high glucose
Products
Product Category
Brand
Physical Form
Glucose (g/L)
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing