Skip to Content
Merck

Skip To

CE0050

CelLytic MEM Protein Extraction Kit

Synonym(s):

Membrane protein extraction kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
41116134
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

protein extraction: suitable

Quality Segment

shipped in

wet ice

storage temp.

−20°C

General description

CelLytic MEM Protein Extraction Kit offers a fast and convenient method to isolate hydrophobic and raft microdomain associated proteins from cells. The method is based on phase separation and does not require cell membrane isolation. The separated proteins can be used for further experiments such as SDS-PAGE, Western blotting, dot blotting, and immunoprecipitation. The kit has been tested on, but not limited to, HeLa, HEK-293, NIH 3T3, COS and CHO cell lines.
Membrane proteins make up around 20-30% of an organism′s genome and serve as cellular gatekeepers, regulators, and sensors. They have diverse cellular functions, such as shielding the cell from external toxins, being the starting point of intracellular signaling cascades, and retaining critical ion concentrations.[1]

Application

CelLytic MEM Protein Extraction Kit has been used for de novo lipogenesis measurements using hepatocytes[2] and to extract membrane proteins for western blotting.[3][4]
Sufficient reagents supplied for 80 tests.

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CB0500C2978C1990
technique(s)

protein extraction: suitable

technique(s)

protein extraction: suitable

technique(s)

-

technique(s)

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

wet ice

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C


Kit Components Only

Product No.
Description

  • Lysis and Separation Buffer 50 mL

  • Wash Buffer for CelLytic MEM 50 mL

  • Sodium Chloride, 4M Solution 1.5 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution 1 mL
    SDS

pictograms

Exclamation mark

Storage Class

10 - Combustible liquids

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

wgk

WGK 2

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters