Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.56
UNSPSC Code:
41116134
Pricing and availability is not currently available.
General description
CelLytic™ Express is a non-denaturing and highly efficient protein extraction formulation for in-culture bacterial cell lysis. This proprietary powder formulation extracts 2-3 times more protein than conventional methods such as sonication. In addition, the method is fast and requires less sample manipulations, reducing proteolytic degradation and preserving protein activity. Unlike other in-culture lysis products, CelLytic™ Express is a complete formulation including lysozyme and endonuclease. The resulting lysate is completely clear of cellular debris and is immediately ready for affinity purification. CelLytic™ Express unique powder formulation adds minimal volume to the final lysate. It provides a cell lysis method with defined volume and time parameters, making it easier to validate and more convenient for production scale protocols.
Application
CelLytic Express is a non-denaturing and highly efficient protein extraction formulation for in-culture bacterial cell lysis. This proprietary powder formulation extracts 2-3 times more protein than conventional methods such as sonication. In addition, the method is fast and requires less sample manipulations, reducing proteolytic degradation and preserving protein activity. Unlike other in-culture lysis products, CelLytic Express is a complete formulation including lysozyme and endonuclease. The resulting lysate is completely clear of cellular debris, and is immediately ready for affinity purification. CelLytic Express′ unique powder formulation adds minimal volume to the final lysate. It provides a cell lysis method with defined volume and time parameters, making it easier to validate and more convenient for production scale protocols.
CelLytic™ Express has been used as a component in lysis buffer for protein purification.[1]
CelLytic Express is optimized for the lysis of E. coli strain BL21. However, it works well with other commons strains such as JM109. CelLytic Express may also be used on other similar bacterial cells. CelLytic Express has been tested with BL21 E. coli cells expressing histidine-tagged and FLAG fusion proteins. It should be compatib with affinity purification of other fusion proteins.
CelLytic Express is optimized for the lysis of E. coli strain BL21. However, it works well with other commons strains such as JM109. CelLytic Express may also be used on other similar bacterial cells. CelLytic Express has been tested with BL21 E. coli cells expressing histidine-tagged and FLAG fusion proteins. It should be compatib with affinity purification of other fusion proteins.
Features and Benefits
- Efficient and non-denaturing in-culture protein extraction
- Save time by eliminating cell harvest and clarification steps
- Preserve biological activity with fewer manipulations
- Compatible with affinity resins and magnetic beads
- Ideal for Production Scale Extraction protocols
Preparation Note
CelLytic™ Express is supplied ready-to-use. However, additional reagents may be used depending on the nature of the protein to be isolated. These include salts, protease inhibitors, and reducing agents such as dithiothreitol or 2-mercaptoethanol. The additional reagents should be added into the bacterial culture after dissolving the CelLytic™ Express, and not by adding them to the dry powder.
This product ships on wet ice and storage at –20°C is recommended. CelLytic™ Express, as supplied, is stable for at least two years when stored properly.
Other Notes
One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.
Legal Information
CelLytic is a trademark of Sigma-Aldrich Co. LLC
1 of 1
This Item | |||
|---|---|---|---|
| shipped in wet ice | shipped in - | shipped in wet ice | shipped in - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form powder | form solution | form - | form solution |
| storage temp. −20°C | storage temp. - | storage temp. −20°C | storage temp. room temp |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Veronica Nasta et al.
The FEBS journal, 287(11), 2312-2327 (2019-11-15)
During its late steps, the mitochondrial iron-sulfur cluster (ISC) assembly machinery leads to the formation of [4Fe-4S] clusters. In vivo studies revealed that several proteins are implicated in the biosynthesis and trafficking of [4Fe-4S] clusters in mitochondria. However, they do not
Emmanuelle Szenker et al.
Cell reports, 1(6), 730-740 (2012-07-21)
Discovering how histone variants that mark distinct chromatin regions affect a developmental program is a major challenge in the epigenetics field. To assess the importance of the H3.3 histone variant and its dedicated histone chaperone HIRA, we used an established
Jiaquan Liu et al.
Journal of molecular biology, 430(22), 4456-4468 (2018-06-05)
DNA mismatch repair (MMR) is a DNA excision-resynthesis process that principally enhances replication fidelity. Highly conserved MutS (MSH) and MutL (MLH/PMS) homologs initiate MMR and in higher eukaryotes act as DNA damage sensors that can trigger apoptosis. MSH proteins recognize
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C1990-25ML | 04061835506125 |
| C1990-6X500ML | 04061835572205 |
| C1990-10X25ML | 04061835506118 |
| C1990-500ML | 04061838247476 |




