Skip to Content
Merck

Skip To

C7521

Human Collagen Type IV

from human placenta, powder, suitable for substrate for collagenase and for diagnostic assay manufacturing

Synonym(s):

Collagen Type 4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.54
EC Number:
232-697-4
MDL number:
Biological source:
human placenta
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Collagen from human placenta, Bornstein and Traub Type IV, powder

biological source

human placenta

Quality Segment

description

Sigma Type VI

form

powder

impurities

HIV, hepatitis B and hepatitis C, none detected

solubility

0.5 M acetic acid: 1 mg/mL

suitability

suitable for substrate for collagenase

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Biochem/physiol Actions

During development, collagen IV is ubiquitously distributed in BMs. During the maturation process, this network gets partially replaced in a remarkably tissue specific manner, defining BM structure and function. Collagen IV has been shown to bind to platelets, hepatocytes, keratinocytes, endothelial, mesangial, pancreatic cells and some tumor cells.

Preparation Note

This is a lyophilized powder that can be reconstituted in sterile .5 M acetic acid, PBS or water at 1 mg/mL. A PBS solution will be stable for at least one year at -20°C. It is prepared by a modification of the pepsin extraction method of Glanville, R.W., et al.[1]

Analysis Note

An SDS polyacrylamide gel electrophoresis test run under reducing conditions consistent with basement membrane collagen yields three major bands.

Other Notes

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue. Type IV collagen occurs only in the basement membranes and contains up to six genetically distinct a-chains.
Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Be wary of confusing Sigma-type designations with recognized collagen classification types.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C3657C7774C4407
biological source

human placenta

biological source

human placenta

biological source

human placenta

biological source

human placenta

Gene Information

human ... COL4A2(1284), COL4A3(1285), COL4A4(1286), COL4A5(1287), COL4A6(1288)

Gene Information

human ... COL5A1(1289)

Gene Information

bovine ... CSN2(281099), CSN3(281728)
human ... COL4A3(1285), COL4A4(1286), COL4A6(1288)

Gene Information

human ... COL3A1(1300)

form

powder

form

powder

form

powder

form

powder

suitability

suitable for substrate for collagenase

suitability

-

suitability

-

suitability

-

application(s)

diagnostic assay manufacturing

application(s)

detection

application(s)

-

application(s)

-

solubility

0.5 M acetic acid: 1 mg/mL

solubility

aqueous acid: soluble

solubility

-

solubility

aqueous acid: soluble


Still not finding the right product?

Explore all of our products under Collagen from human placenta


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters