Skip to Content
Merck

Skip To

C3657

Human Collagen Type IX

from human placenta, powder, suitable for detection assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32
EC Number:
232-697-4
MDL number:
Form:
powder
Biological source:
human placenta
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder

biological source

human placenta

form

powder

impurities

HIV, hepatitis B and hepatitis C, none detected

solubility

aqueous acid: soluble

UniProt accession no.

application(s)

detection

storage temp.

2-8°C

Gene Information

human ... COL5A1(1289)

Preparation Note

This product was prepared by a modification of the pepsin extraction and salt fractionation method of Niyibizi, C., et al., J. Biol. Chem., 259, 14170 (1984). It is an acid soluble collagen that can be dissolved in water with acetic acid added to pH 3.0 (5 mg/mL), yielding an opalescent, colorless solution.

Analysis Note

An SDS polyacrylamide gel electrophoresis test run under reducing conditions consistent with basement membrane collagen yields three major bands.

Other Notes

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue.
Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Be wary of confusing Sigma-type designations with recognized collagen classification types.

Disclaimer

This product should be stored desiccated at -20°C, and will retain activity in these conditions for 3 years.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7521H4417234149
biological source

human placenta

biological source

human placenta

biological source

human placenta

biological source

-

form

powder

form

powder

form

solution

form

solution

UniProt accession no.

P20908

UniProt accession no.

P08572, P29400, P53420, Q01955, Q14031

UniProt accession no.

P02462, P08572, P29400, P53420, Q01955, Q14031

UniProt accession no.

-

Gene Information

human ... COL5A1(1289)

Gene Information

human ... COL4A2(1284), COL4A3(1285), COL4A4(1286), COL4A5(1287), COL4A6(1288)

Gene Information

human ... COL4A1(1282), COL4A2(1284), COL4A3(1285), COL4A4(1286), COL4A5(1287), COL4A6(1288)

Gene Information

-

impurities

HIV, hepatitis B and hepatitis C, none detected

impurities

HIV, hepatitis B and hepatitis C, none detected

impurities

Endotoxin, tested, HIV, hepatitis B and hepatitis C, none detected

impurities

-

solubility

aqueous acid: soluble

solubility

0.5 M acetic acid: 1 mg/mL

solubility

-

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters