Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-576-6
MDL number:
Pricing and availability is not currently available.
Quality Segment
form
powder
packaging
vial of ~1 mg protein
pI
5.4
solubility
deionized water: soluble, clear to slightly hazy, colorless (Dissolve vial contents in 0.25mL of deionised water.)
storage temp.
−20°C
Preparation Note
Dissolve in 0.25mL of deionized water. The solution is colorless, clear to slightly hazy.
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C |
| solubility deionized water: soluble, clear to slightly hazy, colorless (Dissolve vial contents in 0.25mL of deionised water.) | solubility - | solubility - | solubility - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| packaging vial of ~1 mg protein | packaging - | packaging vial of 5 mg | packaging - |
| pI 5.4 | pI 6.6 | pI - | pI - |
Still not finding the right product?
Explore all of our products under Carbonic Anhydrase Isozyme II from bovine erythrocytes
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




