Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbonic Anhydrase Isozyme II from bovine erythrocytes
Synonym(s):
Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase II
CAS Number:
9001-03-0
EC Number:
232-576-6
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
pK Value
Tissue
Quality Segment
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C2522
Carbonic Anhydrase Isozyme II from bovine erythrocytes,
lyophilized powder, ≥3,000 W-A units/mg protein
Expand
View Pricing
C3666
Carbonic Anhydrase Isozyme II from bovine erythrocytes,
Isoelectric focusing marker, pI 5.4
Expand
View Pricing
C3640
Carbonic Anhydrase Isozyme II from bovine erythrocytes,
lyophilized powder, ≥2,000 W-A units/mg protein
Expand
View Pricing