Skip to Content
Merck

Skip To

C4482

CelLytic Y Cell Lysis Reagent

For yeast cells

Synonym(s):

Yeast cell lysis reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
41116134
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solution

input

bacteria and yeast sample(s)

General description

CelLytic Y Cell Lysis Reagent is versatile, phosphate-free and non-denaturing. It provides an effective method for cell lysis and protein solubilization. Target proteins maintain immunoreactivity or biological function. The protocol is brief and preformed at room temperature. No extreme conditions or glass beads are required. Use only 2.5-5 ml/gram yeast cells.

Application

CelLytic Y Cell Lysis Reagent has been used in lysis of yeast cells expressing DNA repair, Rad protein, heat shock protein (ssa1) and human interleukin 4 (hIL4).

Features and Benefits

  • Fast: Cell lysis in just 20 minutes
  • Efficient: Three times more efficient than glass beads
  • Flexible: Lysis is enhanced by the addition of reducing reagent (DTT)
  • Gentle: Non-denaturing - preserves biological activity

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C1990B7310C8740
description

For yeast cells

description

-

description

For bacterial cell lysis, 2× concentrate

description

For bacterial cell lysis, 10× concentrate

input

bacteria and yeast sample(s)

input

-

input

-

input

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solution

form

powder

form

liquid

form

liquid


pictograms

Exclamation mark

signalword

Danger

Storage Class

10 - Combustible liquids

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–7 of 7 Questions  
  1. Does this contain EDTA or other chelators?

    1 answer
    1. This product does not contain EDTA or other chelators.

      Helpful?

  2. Can I use CelLytic™ Y Cell Lysis Reagent, Product C4482, to make spheroplasts?

    1 answer
    1. No.  For spheroplast formation and protein extraction we recommend the CelLytic Y Plus kit.

      Helpful?

  3. Should I use CelLytic™ Y Cell Lysis Reagent, Product C4482?

    1 answer
    1. For all cases in which the addition of DTT does not interfere with the application tested, it is recommended to add DTT (5 - 10 mM final concentration) to the CelLytic Y reagent.  Addition of DTT significantly improves total protein yield.

      Helpful?

  4. What is in CelLytic™ Y Cell Lysis Reagent, Product C4482?

    1 answer
    1. The formulation is considered proprietary, but we can state that it is a non-denaturing phosphate-free reagent.

      Helpful?

  5. Can I use glass beads with CelLytic™ Y Cell Lysis Reagent, Product C4482?

    1 answer
    1. If cell growth conditions are not optimal, then glass beads can be used to increase yields.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. What strains have been used with CelLytic™ Y Cell Lysis Reagent, Product C4482?

    1 answer
    1. The efficiency of CelLytic Y reagent for protein extraction has been tested on different strains of Saccharomyces cerevisiae [Y187, Y190, W303(a), S288C, SP1, S1278b (diploid and haploid), BJ2168, and cdc25] and Schizosaccharomyces pombe.

      Helpful?

Reviews

No rating value

Active Filters