Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C10H7BrClNO2
CAS Number:
Molecular Weight:
288.53
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Pricing and availability is not currently available.
Quality Segment
assay
≥99% (TLC)
form
powder
solubility
ethanol: 50 mg/mL, clear, colorless to faintly yellow
storage temp.
−20°C
SMILES string
CC(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12
InChI
1S/C10H7BrClNO2/c1-5(14)15-8-4-13-7-3-2-6(11)10(12)9(7)8/h2-4,13H,1H3
InChI key
WPWLFFMSSOAORQ-UHFFFAOYSA-N
General description
A histochemical substrate for esterase.
Packaging
Sealed ampule.
Disclaimer
Decomposes in storage with development of dark blue-green color.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99% (TLC) | assay ≥97% | assay ≥98% (TLC) | assay ≥99% (TLC) |
| solubility ethanol: 50 mg/mL, clear, colorless to faintly yellow | solubility DMF: 50 mg/mL, clear, colorless to faintly yellow | solubility DMF: 50 mg/mL, clear, colorless to faintly yellow | solubility pyridine: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form powder | form powder | form powder | form powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B4377-25MG | 04061833267769 |
| B4377-500MG | 04061833267776 |



