Skip to Content
Merck
All Photos(3)

Key Documents

B7877

Sigma-Aldrich

6-Bromo-2-naphthyl β-D-glucopyranoside

≥99% (TLC)

Synonym(s):

6-Bromo-2-naphthyl β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17BrO6
CAS Number:
Molecular Weight:
385.21
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥99% (TLC)

form

powder

solubility

pyridine: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3cc(Br)ccc3c2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C16H17BrO6/c17-10-3-1-9-6-11(4-2-8(9)5-10)22-16-15(21)14(20)13(19)12(7-18)23-16/h1-6,12-16,18-21H,7H2/t12-,13-,14+,15-,16-/m1/s1

InChI key

NLRXQZJJCPRATR-IBEHDNSVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Bromo-2-naphthyl β-D-glucopyranoside may be used as a substrate for β-glycosidases in fast blue BB assay. It is also suitable for use with fast blue BB reagent in native polyacrylamide gel electrophoresis (PAGE) for β-glycosidases activity staining.

Biochem/physiol Actions

6-Bromo-2-naphthyl β-D-glucopyranoside serves as a substrate for β-glycosidases. When hydrolyzed it forms an insoluble colored product.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Sánchez-Pérez et al.
The Plant journal : for cell and molecular biology, 60(5), 894-906 (2009-08-18)
Traditional methods to localize beta-glycosidase activity in tissue sections have been based on incubation with the general substrate 6-bromo-2-naphthyl-beta-d-glucopyranoside. When hydrolysed in the presence of salt zinc compounds, 6-bromo-2-naphthyl-beta-d-glucopyranoside affords the formation of an insoluble coloured product. This technique does
I Polacheck et al.
Journal of clinical microbiology, 25(5), 907-910 (1987-05-01)
In this report we attempt to explain the discrepancy between beta-glucosidase (EC 3.2.1.21) activity in Candida albicans as measured by commercial kits and that found in an experimental assay. beta-Glucosidase activity in American and Israeli isolates of C. albicans was
Eric Odoux et al.
Journal of agricultural and food chemistry, 51(10), 3168-3173 (2003-05-02)
Vanilla bean beta-D-glucosidase was purified to apparent homogeneity by successive anion exchange, hydrophobic interaction, and size-exclusion chromatography. The enzyme is a tetramer (201 kDa) made up of four identical subunits (50 kDa). The optimum pH was 6.5, and the optimum
Raquel Sánchez-Pérez et al.
The Plant journal : for cell and molecular biology, 60(5), 894-906 (2009-08-18)
Traditional methods to localize beta-glycosidase activity in tissue sections have been based on incubation with the general substrate 6-bromo-2-naphthyl-beta-d-glucopyranoside. When hydrolysed in the presence of salt zinc compounds, 6-bromo-2-naphthyl-beta-d-glucopyranoside affords the formation of an insoluble coloured product. This technique does
A specific stain for alpha-glucosidases in isoelectric focusing gels.
L L Spielman et al.
Analytical biochemistry, 120(1), 66-70 (1982-02-01)

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service