Skip to Content
Merck

Skip To

A9099

Acrylamide

Molecular Biology, ≥99% (HPLC)

Synonym(s):

2-Propenamide, Polyacrylamide, acrylic amide, akrylamid, Acrylic acid amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
NACRES:
NA.31
PubChem Substance ID:
eCl@ss:
39031205
UNSPSC Code:
41105319
EC Number:
201-173-7
MDL number:
Beilstein/REAXYS Number:
605349
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

vapor density

2.45 (vs air)

vapor pressure

0.03 mmHg ( 40 °C)

assay

≥99% (HPLC)

form

crystals

impurities

≤0.005% Acrylic acid content, ≤0.005% Water insolubles

conductivity

≤10 μmho (35% solution)

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

solubility

water: soluble 2.5 g/10 mL, clear, colorless

cation traces

Fe: ≤1 ppm, Mg: ≤3 ppm, heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, RNase, protease, none detected

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

General description

Polyacrylamide gels are prepared using acrylamide and bis-acrylamide. Acrylamide is a water-soluble monomer, which forms a transparent stable insoluble gel. Bis-acrylamide is a cross-linking agent.

Application

Acrylamide has been used:
  • in polyacrylamide gel electrophoresis
  • in the preparation of cleared gel samples
  • to determine the effect of L-cysteine on acrylamide-induced testicular toxicity


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3553A888701696
form

crystals

form

powder

form

crystals

form

powder blend

grade

Molecular Biology

grade

-

grade

-

grade

Molecular Biology

foreign activity

DNase, RNase, protease, none detected

foreign activity

-

foreign activity

-

foreign activity

-

impurities

≤0.005% Acrylic acid content, ≤0.005% Water insolubles

impurities

≤0.005% Acrylic acid, ≤0.005% Insoluble in water

impurities

-

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

assay

≥99% (HPLC)

assay

≥99% (HPLC)

assay

≥99%

assay

≥99.5% (GC)

cation traces

Fe: ≤1 ppm, heavy metals (as Pb): ≤5 ppm, Mg: ≤3 ppm

cation traces

Fe: ≤1 ppm, Mg: ≤3 ppm, heavy metals (as Pb): ≤5 ppm

cation traces

-

cation traces

-


Still not finding the right product?

Explore all of our products under Acrylamide


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A9099-500G04061835542666
A9099-1KG04061833401354
A9099-25G04061835542659
A9099-100G04061835542635
A9099-2.5KG04061835542642

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters