Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acrylamide
Synonym(s):
2-Propenamide, Acrylic acid amide, Acrylamide
Linear Formula:
CH
2
=CHCONH
2
CAS Number:
79-06-1
Molecular Weight:
71.08
EC Number:
201-173-7
Products
Brand
Millipore (1)
Sigma-Aldrich (6)
Supelco (2)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
certified reference material (1)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
A8887
Acrylamide,
suitable for electrophoresis, ≥99%
Expand
View Pricing
A9099
Acrylamide,
Molecular Biology, ≥99% (HPLC)
Expand
View Pricing
8.00830
Acrylamide,
for synthesis
Expand
View Pricing
A3553
Acrylamide,
suitable for electrophoresis, ≥99% (HPLC), powder
Expand
View Pricing
1.10784
Acrylamide,
for electrophoresis
Expand
View Pricing
23701
Acrylamide,
analytical standard
Expand
View Pricing
01700
Acrylamide,
purum, ≥98.0% (GC)
Expand
View Pricing
08267
Acrylamide,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
01696
Acrylamide,
for Northern and Southern blotting, powder blend
Expand
View Pricing