Skip to Content
Merck
All Photos(1)

Documents

A6770

Sigma-Aldrich

Methotrexate hydrate

powder, ≥98% (HPLC)

Synonym(s):

4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Methotrexate hydrate, ≥98% (HPLC), powder

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

color

, yellow to very dark yellow to very dark orange

mp

185-204  °C

solubility

H2O: insoluble

storage temp.

−20°C

SMILES string

[H]O[H].CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5.H2O/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);1H2/t13-;/m0./s1

InChI key

FPJYMUQSRFJSEW-ZOWNYOTGSA-N

Gene Information

human ... DHFR(1719)

Looking for similar products? Visit Product Comparison Guide

Application

Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also shows immunosuppressive effects in, e.g., rheumatoid arthritis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bokyeong Ryu et al.
Frontiers in veterinary science, 8, 587659-587659 (2021-10-05)
The gastrointestinal tract is the most common exposure route of xenobiotics, and intestinal toxicity can result in systemic toxicity in most cases. It is important to develop intestinal toxicity assays mimicking the human system; thus, stem cells are rapidly being
Methotrexate: new uses for an old drug.
Philip J Hashkes et al.
The Journal of pediatrics, 164(2), 231-236 (2013-11-30)
Yingjuan J Lu et al.
Cell reports. Medicine, 2(10), 100422-100422 (2021-11-11)
Provoked by sterile/nonsterile insults, prolonged monocyte mobilization and uncontrolled monocyte/macrophage activation can pose imminent or impending harm to the affected organs. Curiously, folate receptor beta (FRβ), with subnanomolar affinity for the vitamin folic acid (FA), is upregulated during immune activation
Josef Singer et al.
Molecular cancer therapeutics, 13(7), 1777-1790 (2014-04-24)
Passive immunotherapy with monoclonal antibodies represents a cornerstone of human anticancer therapies, but has not been established in veterinary medicine yet. As the tumor-associated antigen EGFR (ErbB-1) is highly conserved between humans and dogs, and considering the effectiveness of the
Behnoush Hajian et al.
Cell chemical biology, 26(6), 781-791 (2019-04-02)
The folate biosynthetic pathway offers many druggable targets that have yet to be exploited in tuberculosis therapy. Herein, we have identified a series of small molecules that interrupt Mycobacterium tuberculosis (Mtb) folate metabolism by dual targeting of dihydrofolate reductase (DHFR), a

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service