Skip to Content

M7824

Methotrexate

Synonym(s):

Methotrexate, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylpteroyl-L-glutamic acid, Antifolan, MTX, Methylaminopterin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44
EC Number:
200-413-8
UNSPSC Code:
41116107
NACRES:
NA.21
Beilstein/REAXYS Number:
70669
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

form

powder

suitability

suitable for manufacturing use

application(s)

downstream processes
pharma/biopharma processes

storage temp.

−20°C

SMILES string

CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1

InChI key

FBOZXECLQNJBKD-ZDUSSCGKSA-N

Gene Information

human ... DHFR(1719)

General description

Inhibits thymidylate synthetase, is a nonselective de novo purine synthesis inhibitor and has a significant toxicity profile including hepatotoxicity, pneumonitis and bone marrow suppression. Potent folic acid antagonist. Induces apoptosis in HL-60 human leukemia cells. Also useful as an anti-tumor agent.
Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Methotrexate


Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 1B - STOT RE 1

Target Organs

Liver,Bone marrow

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library