Skip to Content
Merck
All Photos(4)

Key Documents

A1296

Sigma-Aldrich

Agar

microbiology tested, suitable for plant cell culture, suitable for cell culture, powder

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
eCl@ss:
42040102
NACRES:
NA.21

biological source

algae (Rhodophyceae)

form

powder

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable
microbiological culture: suitable

transition temp

congealing temperature <38 °C (1.5% in H2O)

suitability

microbiology tested

application(s)

agriculture
agriculture
microbiology
microbiology

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Agar is commonly employed in research for:
  • agar plates for microbiology, such as Tryptic soy broth or TSB Agar plates for bacterial cell culture
  • 50% Murashige-Skoog or MS medium (basal MS)for growing plant seedlings
  • micropropagation of Pistacia lentiscus or Lentisk, for shoot initiation

Features and Benefits

  • High-quality agar powder for general use in research
  • Suitable for microbiology applications to culture bacteria and other microorganisms
  • Suitable for plant cell culture for horticultural science and biotechnology
  • Easily prepare and autoclave for a solid or semi-solid media at 6 - 12g/L

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inhibited growth of Pseudomonas aeruginosa by dextran- and polyacrylic acid-coated ceria nanoparticles
Wang Q et al.
International journal of nanomedicine, 8, 3395-3399 (2013)
Mineko Konishi et al.
BMC plant biology, 19(1), 90-90 (2019-03-02)
NIN-LIKE PROTEIN (NLP) transcription factors are master regulators of nitrate-inducible gene expression in higher plants. NLP transcription factors contain a nitrate signal-responsive domain in the amino-terminal region, an RWP-RK-type DNA-binding domain in the middle, and a Phox and Bem1 (PB1)
Plant regeneration from callus derived from immature embryo cotyledons of Prunus mume.
Ning GG and Bao MZ
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 42, 744-747 (2007)
Micropropagation of mature Quercus ilex L. trees by axillary budding
Martinez MT et al.
Plant Cell, Tissue and Organ Culture, 131, 499?512-499?512 (2017)
Essential role of the CD docking motif of MPK4 in plant immunity, growth, and development
Siodmak A et al.
The New phytologist, 239, 1112-1126 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service