Skip to Content

A4800

Agar

Type M, suitable for plant cell culture

Synonym(s):

Agar-agar, Gum agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
UNSPSC Code:
50405901
eCl@ss:
42040102
EC Number:
232-658-1
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (Rhodophyceae)

Quality Segment

type

Type M

form

powder

technique(s)

cell culture | plant: suitable

transition temp

transition temp 34-36 °C

application(s)

agriculture

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

General description

Type M agar is predominantly used in plant tissue culture. It is similar to agar, a polysaccharide complex with a backbone of alternating agarose and agaropectin molecules and extracted from red algae.

Application

Type M agar has been used as a component of Murashige and Skoog (MS) medium for culturing Arabidopsis thalania seeds. It has also been used in rooting media for tomato (Solanum lycopersicum) seeds.


Still not finding the right product?

Explore all of our products under Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library