Skip to Content
Merck

Skip To

914

Sodium nitrite solution

Synonym(s):

Sodium nitrite preparation

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Form:
solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

100 mmol/L

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Application

For use in the Naphthol AS-D Chloroacetate Esterase and Naphthyl Acetate Esterase Procedures, Kits 91C and 91A.

Other Notes

0.1 mol/L

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
72586521051563218
form

solution

form

solution

form

liquid

form

solid

Quality Level

500

Quality Level

200

Quality Level

100

Quality Level

200

application(s)

hematology
histology

application(s)

-

application(s)

-

application(s)

-

IVD

for in vitro diagnostic use

IVD

-

IVD

-

IVD

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

concentration

100 mmol/L

concentration

0.1 M NO2-

concentration

27-33% (titration by KMnO4), 30 wt. % in H2O

concentration

-


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tiffany A Major et al.
Antimicrobial agents and chemotherapy, 54(11), 4671-4677 (2010-08-11)
A hallmark of airways in patients with cystic fibrosis (CF) is highly refractory, chronic infections by several opportunistic bacterial pathogens. A recent study demonstrated that acidified sodium nitrite (A-NO(2)(-)) killed the highly refractory mucoid form of Pseudomonas aeruginosa, a pathogen
Paul J Shami et al.
Journal of medicinal chemistry, 49(14), 4356-4366 (2006-07-11)
The literature provides evidence that metabolic nitric oxide (NO) release mediates the cytotoxic activities (against human leukemia and prostate cancer xenografts in mice) of JS-K, a compound of structure R(2)N-N(O)=NO-Ar for which R(2)N is 4-(ethoxycarbonyl)piperazin-1-yl and Ar is 2,4-dinitrophenyl. Here
Deniz Ekinci et al.
Bioorganic & medicinal chemistry, 19(2), 744-748 (2011-01-08)
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of



Global Trade Item Number

SKUGTIN
PH015305-1G04061825646336
CDS022231-25MG04061829028275
PH015123-100MG04061831271690
CDS021975-100MG04061829017880
PH015396-1G04061825647135
CDS023040-25MG04061829010867
67575-100MG04061832736143
111961-100G04061838699336
111961-500G04061838699343
914-10ML04061833263075
460125-100ML04061837812590
GF52616875-1EA04061825096353
GF25475272-1EA04061825086460
GF30365917-1EA04061825092362
GF32571806-1EA04061825092676
GF38981065-1EA04061825094083
GF47160738-1EA04061825095431
GF68647977-1EA04061825088365
GF91538196-1EA04061825099798
GF87852505-1EA04061825090368
GF98203552-1EA04061825091778

Questions

Reviews

No rating value

Active Filters