Skip to Content
Merck
All Photos(2)

Key Documents

521051

Sigma-Aldrich

Calcium nitrite solution

30 wt. % in H2O

Synonym(s):

Calcium dinitrite, Calcium nitrite, Calcium nitrite (Ca(NO2 )2 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO2)2
CAS Number:
Molecular Weight:
132.09
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid

concentration

27-33% (titration by KMnO4)
30 wt. % in H2O

refractive index

n20/D 1.3946

density

1.30 g/mL

SMILES string

[Ca++].[O-]N=O.[O-]N=O

InChI

1S/Ca.2HNO2/c;2*2-1-3/h;2*(H,2,3)/q+2;;/p-2

InChI key

AONJRPXZCVADKF-UHFFFAOYSA-L

General description

Calcium nitrite solution (Ca(NO2)2) is widely utilized as a corrosion inhibitor. It protects the metal surface against chloride attack and increases the threshold level and improves the overall life of the concrete. Ca(NO2)2 does not change the mechanical properties of the cement.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel approach for quantitative LIBS fluorine analysis using CaF emission in calcium-free samples
Alvarez-Llamas C, et al.
Journal of Analytical Atomic Spectrometry, 32(1), 162-166 (2017)
Xuan Li et al.
Environmental science & technology, 54(4), 2323-2333 (2020-01-25)
Microbially induced concrete corrosion is a major deterioration process in sewers, causing a huge economic burden, and improved mitigating technologies are required. This study reports a novel and promising effective solution to attenuate the corrosion in sewers using calcium nitrite-admixed
Rosemary M Onjiko et al.
Analytical chemistry, 89(13), 7069-7076 (2017-04-25)
Knowledge of single-cell metabolism would provide a powerful look into cell activity changes as cells differentiate to all the tissues of the vertebrate embryo. However, single-cell mass spectrometry technologies have not yet been made compatible with complex three-dimensional changes and
Effect of calcium nitrite-based corrosion inhibitor in preventing corrosion of embedded steel in concrete
Ann K, et al.
Cement and Concrete Research, 36(3), 530-535 (2006)
Rosemary M Onjiko et al.
Analytical methods : advancing methods and applications, 9(34), 4964-4970 (2017-10-25)
Single-cell mass spectrometry (MS) empowers the characterization of metabolomic changes as cells differentiate to different tissues during early embryogenesis. Using whole-cell dissection and capillary electrophoresis electrospray ionization (CE-ESI) MS, we recently uncovered metabolic cell-to-cell differences in the 8- and 16-cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service