Skip to Content
Merck

Skip To

85547

Span® 65

Synonym(s):

Sorbitan tristearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
EC Number:
247-891-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Level

form

solid

technique(s)

protein quantification: suitable

HLB value

2.1±1.0

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)C1OC[C@H](OC(=O)CCCCCCCCCCCCCCCCC)[C@H]1OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C60H114O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-52-54(61)59-60(68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)55(53-66-59)67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h54-55,59-61H,4-53H2,1-3H3/t54-,55+,59-,60-/m1/s1

InChI key

IDGUCWCSGWRLPK-LLPUSWRMSA-N

General description

Span® 65 is an oil-soluble surfactant. It is used as an emulsifier, stabilizer, and food additive (E number E492). Span 65 emulsifier is broadly used for its capability to alter the crystal morphology and consistency of fats. It can be used as an anti-bloom agent in confectionery products comprising cocoa butter and cocoa butter substitutes. Hence, it acts as a possible controller of crystallization. Span 65 effectively stabilizes the polymorph β′ of margarine and changes the solid fat content (SFC). This boosts fusion profiles appropriate for the body temperature. Span 65 also helps in the generation of organogels.

Application

Span® 65 has been used as a plasticizer candidate to study the plasticization, brittleness, and loading curve characteristics of pure carnauba wax. It has also been used as an emulsifying agent to prepare flaxseed oil emulsions.

Legal Information

Span is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.40122S7135S7010
form

solid

form

powder

form

viscous liquid

form

-

technique(s)

protein quantification: suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

description

non-ionic

description

-

description

non-ionic

description

non-ionic

HLB value

2.1±1.0

HLB value

-

HLB value

-

HLB value

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

302.0 °F - closed cup

flash_point_c

150.00 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



PREPARATION_OF_FLAXSEED_OIL_EMULSIONS
Al-AchiA. and Pranshu S
Journal of Pharmaceutical and Scientific Innovation (2015)
Structural characteristics of crystals formed in palm oil using sorbitan tristearate and sucrose stearate
Domingues MA, et al.
International journal of food properties, 21(1), 618-632 (2018)
Surface rheology of sorbitan tristearate and beta-lactoglobulin: Shear and dilatational behavior
Torcello-Gomez A, et al.
Journal of Non-Newtonian Fluid Mechanics, 166(12-13), 713-722 (2011)



Global Trade Item Number

SKUGTIN
85547-250G04061833025505
85547-1KG04061826271230

Questions

Reviews

No rating value

Active Filters