Skip to Content
Merck
All Photos(3)

Documents

S7010

Sigma-Aldrich

Span® 60

Synonym(s):

Sorbitan stearate, Sorbitane monostearate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

mol wt

430.63 g/mol

concentration

45-55% (GC)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C24H46O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h20-21,23-26,28H,2-19H2,1H3/t20-,21?,23+,24+/m0/s1

InChI key

HVUMOYIDDBPOLL-XGKPLOKHSA-N

General description

Span® 60 is a sorbitan monoester than is used as a non-ionic detergent.

Application

Span® 60 has been used in a study to assess encapsulation of doxorubicin in niosomes as a route to tumor targeting. It has also been used in a study to investigate the use of nonionic surfactants as contrast agents for use in diagnostic ultrasounds.

Other Notes

Fatty acid composition: Stearic acid (C18:0) approx. 50%; balance primarily palmitic acid (C16:0).

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and properties of vesicles (niosomes) of sorbitan monoesters ([TM="Span"] 20, 40, 60 and 80) and a sorbitan triester ([TM="Span"] 85).
Yoshioka, T., et al.
International Journal of Pharmaceutics, 105, 1-1 (1994)
I F Uchegbu et al.
Pharmaceutical research, 12(7), 1019-1024 (1995-07-01)
Encapsulation of doxorubicin in niosomes was sought as a route to tumour targeting and improved tumoricidal through the alteration of doxorubicin pharmacokinetics and metabolism. Doxorubicin niosomes (10 mg kg-1 doxorubicin) prepared from sorbitan monostearate (Span 60), cholesterol and choleth-24 (a
Surfactant-stabilized microbubbles as ultrasound contrast agents: stability study of [TM="Span"] 60 and Tween 80 mixtures using a Langmuir trough
Singhal, S., et al.
Langmuir, 9, 2426-2426 (1993)
Karimunnisa Sameer Shaikh et al.
Drug development and industrial pharmacy, 36(8), 946-953 (2010-03-04)
Niosomal delivery can prove an alternative to improve the poor skin penetration and residence of the topical antifungal drugs that account for the long treatment regimes in cutaneous mycosis. To investigate niosomes as carriers for dermal delivery of ciclopirox olamine
F Podczeck et al.
International journal of pharmaceutics, 361(1-2), 33-40 (2008-06-24)
The aim of the study was to investigate the possibility of incorporating non-ionic surfactants into pellets produced from microcrystalline cellulose by the process of extrusion/spheronization and the properties of the pellets. A hydrophilic surfactant, polysorbate 60 (PS 60), and two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service