Skip to Content
Merck

Skip To

61360

Lactulose

≥98.0% (HPLC)

Synonym(s):

4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
225-027-7
Beilstein/REAXYS Number:
93773
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98.0% (HPLC)

form

solid

optical activity

[α]20/D −48±2°, 24 hr, c = 1% in H2O

color

white to off-white

mp

169  °C ((336 °F ))

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO)O[C@@](O)(CO)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)7(17)8(18)11(21-4)22-9-5(2-14)23-12(20,3-15)10(9)19/h4-11,13-20H,1-3H2/t4-,5-,6+,7+,8-,9-,10+,11+,12+/m1/s1

InChI key

JCQLYHFGKNRPGE-WJONTELPSA-N

General description

Lactulose is a synthetic derivative of lactose. It is made up of one molecule of galactose and one molecule of fructose.

Application

Lactulose has been used in the preparation of milk.
Lactulose, a disaccharide composed of fructose and galactose units, is used as an osmolyte that relieves constipation and supports probiotic processes in the intestine and colon. Lactulose helps stabilize blood glucose and insulin levels.

Biochem/physiol Actions

Lactulose helps to decrease the concentration of blood ammonia in the treatment of hepatic encephalopathy. It is also used to treat constipation as an osmotic laxative. This non-absorbable disaccharide is catabolized through the bacterial flora to short chain fatty acids. The enzymes of the mammalian digestive tract do not have the ability to digest lactulose.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L7877PHR1608BP210
Lactulose British Pharmacopoeia (BP) Reference Standard

BP210

Lactulose

assay

≥98.0% (HPLC)

assay

≥95% (HPLC)

assay

-

assay

-

Quality Level

100

Quality Level

200

Quality Level

300

Quality Level

-

form

solid

form

powder

form

-

form

solid

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

color

white to off-white

color

white to off-white

color

-

color

-

mp

169  °C ((336 °F ))

mp

169  °C ((336 °F ))

mp

-

mp

-


Still not finding the right product?

Explore all of our products under Lactulose


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lactulose enemas in the treatment of hepatic encephalopathy. Do we help or harm?
de Lorenzo-Pinto A, et al.
Revista espa?ola de Enfermedades Digestivas : Organo Oficial de la Sociedad Espa?ola de Patologia Digestiva (2017)
Use of lactulose as prebiotic and its influence on the growth, acidification profile and viable counts of different probiotics in fermented skim milk
Oliveira R P D S, et al.
International Journal of Food Microbiology, 145(1), 22-27 (2011)
Gastrointestinal drugs
Small Animal Clinical Pharmacology, 469-497 (2008)



Global Trade Item Number

SKUGTIN
61360-25G04061832700045
61360-5G04061832700052

Questions

Reviews

No rating value

Active Filters