Skip to Content
Merck
All Photos(2)

Key Documents

L7877

Sigma-Aldrich

Lactulose

≥95% (HPLC)

Synonym(s):

4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93773
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]20/D -52.0 to -48.0 °, c = 4% (w/v) in water

color

white to off-white

mp

169  °C ((336 °F ))

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO)O[C@@](O)(CO)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)7(17)8(18)11(21-4)22-9-5(2-14)23-12(20,3-15)10(9)19/h4-11,13-20H,1-3H2/t4-,5-,6+,7+,8-,9-,10+,11+,12+/m1/s1

InChI key

JCQLYHFGKNRPGE-WJONTELPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lactulose is a synthetic disaccharide derived from galactose and fructose. It is not absorbed in the gut and is frequently used in the treatment of constipation and hepatic encephalopathy. Studies have reported that lactulose administration may cause an increase in fecal nitrogen. It is also known to function as a prebiotic. Lactulose measurements can be used to detect gastrointestinal dysfunctions.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huifang Yin et al.
Carbohydrate research, 465, 58-65 (2018-06-27)
Oligosaccharides derived from lactulose (β-D-Galp-(1 → 4)-D-Fru) are drawing more and more attention nowadays because of their strong resistance to gut digestion, and the interest to discover novel prebiotics. Compared to galactooligosaccharides, currently known structures of lactulose oligosaccharides are very limited. In
Tara Raftery et al.
United European gastroenterology journal, 3(3), 294-302 (2015-07-03)
Vitamin D (vitD) supplementation may prolong remission in Crohn's disease (CD); however, the clinical efficacy and mechanisms are unclear. To determine changes in intestinal permeability (IP), antimicrobial peptide (AMP) concentrations and disease markers in CD, in response to vitD supplementation.
R Mathur et al.
The Journal of clinical endocrinology and metabolism, 98(4), E698-E702 (2013-03-28)
Colonization of the gastrointestinal tract with methanogenic archaea (methanogens) significantly affects host metabolism and weight gain in animal models, and breath methane is associated with a greater body mass index (BMI) among obese human subjects. The objective of the study
B Le Nevé et al.
The American journal of gastroenterology, 108(5), 786-795 (2013-04-17)
The aim of the present pilot study was to evaluate the usefulness of a test meal containing lactulose in the non-invasive assessment of visceral sensitivity in irritable bowel syndrome (IBS), and to identify subsets of IBS patients based on gastrointestinal
C P Kelly et al.
Alimentary pharmacology & therapeutics, 37(2), 252-262 (2012-11-21)
Coeliac disease, an autoimmune disorder triggered by gluten ingestion, is managed by a gluten-free diet (GFD), which is difficult for many patients. Larazotide acetate is a first-in-class oral peptide that prevents tight junction opening, and may reduce gluten uptake and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service