Skip to Content
Merck

Skip To

21902

Carboxymethylcellulose sodium salt

medium viscosity

Synonym(s):

Sodium carboxymethylcellulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

availability

not available in USA

extent of labeling

0.60-0.95 substitution

loss

≤15% loss on drying, 110 °C

color

light yellow

pH

6.5-8.0 (10 mg/mL in H2O)

viscosity

300-1000 mPa.s, 2 % in H2O(25 °C) (dry substance)

mp

> 300.05  °C ((> 572.09 °F ))

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Application

Soluble carboxymethylcelluloses (CM-cellulose; CMC) available in varying viscosities are used as viscosity modifiers (thickeners) to stabilize emulsions and as a chemical dispersants of oils and other carbon structures such as nanotubes. CMCs are used in the development of biostructures such as biofilms, emulsions and nanoparticles for drug delivery.

Other Notes

Characterization of endoglucanase III from Trichoderma reesei[1].
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C488821904419338
Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

powder

form

powder

color

light yellow

color

light yellow

color

beige

color

-

mp

> 300.05  °C ((> 572.09 °F ))

mp

> 300.05 °C ((> 572.09 °F))

mp

-

mp

270 °C (dec.)

extent of labeling

0.60-0.95 substitution

extent of labeling

-

extent of labeling

-

extent of labeling

0.9 carboxymethyl groups per anhydroglucose unit

availability

not available in USA

availability

-

availability

not available in USA

availability

-


Still not finding the right product?

Explore all of our products under Carboxymethylcellulose sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
21902-100G04061835365418
21902-250G04061838775764
21902-1KG04061835365425

Questions

1–3 of 3 Questions  
  1. What is the molecular weight of this product?

    1 answer
    1. The molecular weight of this product is not determined on a lot-to-lot basis. However, historical data indicates a molecular weight range of 160,000-200,000 daltons.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters