Skip to Content
Merck

Skip To

C4888

Carboxymethylcellulose sodium salt

Medium viscosity

Synonym(s):

Sodium carboxymethylcellulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

wood (pulp)

Quality Segment

form

powder

color

light yellow

useful pH range

(20 °C, 10 g/L, 68 °F neutral)

viscosity

400-800 cP, 2 % in H2O(25 °C)(lit.)

mp

> 300.05 °C ((> 572.09 °F))

solubility

water: soluble

cation traces

Na: 6.5-9.5%

application(s)

advanced drug delivery

storage temp.

room temp

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

General description

This product is a medium viscosity carboxymethylcellulose (CMC); the viscosity of a 2% solution in water at 25 C is 400-800 centipoise (cps). The viscosity is both concentration and temperature dependent. As the temperaure increases, the viscosity decreases. As the concentration increases, the viscosity increases.

Low, medium and high viscosity CMCs are all used as suspending agents. Low viscosity CMC is usually used in "thin" aqueous solutions. Medium viscosity CMC is used to make solutions that look like a syrup. High viscosity CMC is used to make a mixture, which resembles a cream or lotion.

Application

Soluble carboxymethylcelluloses (CM-cellulose; CMC) available in varying viscosities are used as viscosity modifiers (thickeners) to stabilize emulsions and as a chemical dispersants of oils and other carbon structures such as nanotubes. CMCs are used in the development of biostructures such as biofilms, emulsions and nanoparticles for drug delivery. Carboxymethylcellulose, medium viscosity, may be used to make solutions the consistency of syrup.[1][2]

Preparation Note

The product is soluble in water (20 mg/ml).
The key to dissolving carboxymethylcellulose is to add the solid carefully to the water so that it is well dispersed (well-wetted). Adding the solid in portions may be necessary. Adding water to the dry solid produces a "clump" of solid that is very difficult to dissolve; the solid must be added to the water. Stir gently or shake intermittently; do not stir constantly with a magnetic stirring bar. High heat is not needed and may actually slow down the solubilization process. A mixing device, such as an impeller-type agitator which produces a vortex, would allow the powder to be drawn into the liquid, but it may produce some shearing.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C5678C501321902
description

Medium viscosity

description

low viscosity

description

High viscosity

description

medium viscosity

biological source

wood (pulp)

biological source

synthetic (organic)

biological source

wood (pulp)

biological source

-

solubility

water: soluble

solubility

water: soluble

solubility

H2O: 10 mg/mL

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

-

cation traces

Na: 6.5-9.5%

cation traces

Na: 6.5-9.5%

cation traces

Na: 6.5-9.5%

cation traces

-


Still not finding the right product?

Explore all of our products under Carboxymethylcellulose sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. what is the average Mw of this product?

    1 answer
    1. The molecular weight is not determined for this product. Historical data shows an estimated range of 100 - 250 kDa.

      Helpful?

Reviews

No rating value

Active Filters