Skip to Content
Merck

Skip To

01673

Acriflavine

BioReagent, suitable for fluorescence, ≥90% (AT)

Synonym(s):

3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H14ClN3
CAS Number:
Molecular Weight:
259.73
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Level

assay

≥90% (AT)

form

powder

composition

Sum of components 3,6-diaminoacridinium chloride and 3,6-diamino-10-methylacridinium chloride, ≥70% HPLC

solubility

H2O: 0.33 g/mL (lit.)(lit.)

fluorescence

λex 463 nm; λem 490 nm in ethanol

suitability

suitable for fluorescence

SMILES string

[Cl-].Nc1ccc2cc3ccc(N)cc3nc2c1.C[n+]4c5cc(N)ccc5cc6ccc(N)cc46

InChI

1S/C14H13N3.C13H11N3.ClH/c1-17-13-7-11(15)4-2-9(13)6-10-3-5-12(16)8-14(10)17;14-10-3-1-8-5-9-2-4-11(15)7-13(9)16-12(8)6-10;/h2-8H,1H3,(H3,15,16);1-7H,14-15H2;1H

InChI key

PEJLNXHANOHNSU-UHFFFAOYSA-N

Application

Acriflavine is used to fluorescently label high molecular weight RNA molecules .
Excitation and emission wavelengths in various solvents :
  • Methanol: λex = 424 nm; λem = 518 nm
  • Ethanol: λex = 426 nm; λem = 524 nm
  • Propanol: λex = 430 nm; λem = 512 nm
  • Butanol: λex = 430 nm; λem = 526 nm
  • Formamide: λex = 434 nm; λem = 524 nm
  • Glycerol: λex = 432 nm; λem = 540 nm
  • Water: λex = 416 nm; λem = 514 nm

Insoluble in ether, chloroform, and fixed oils. Utilized in fluorescence steady state measurements as a donor molecule (when paired with rhodamine 6G as the acceptor) to function as a pH sensor .

Other Notes

DNA fluorochrome. Application in dual fluorescence analysis of cellular DNA and protein simultaneously[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8126A8251SML3350
suitability

suitable for fluorescence

suitability

-

suitability

-

suitability

-

fluorescence

λex 463 nm; λem 490 nm in ethanol

fluorescence

-

fluorescence

-

fluorescence

-

form

powder

form

powder

form

powder

form

powder

solubility

H2O: 0.33 g/mL (lit.)(lit.)

solubility

H2O: 0.33 g/mL (lit.)(lit.)

solubility

H2O: 1 mg/mL

solubility

DMSO: 2 mg/mL, clear

composition

Sum of components 3,6-diaminoacridinium chloride and 3,6-diamino-10-methylacridinium chloride, ≥70% HPLC

composition

-

composition

Acriflavine HCL content, 10-50% , Proflavine HCL, 25-70%

composition

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under Acriflavine


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M Stöhr et al.
Stain technology, 53(4), 205-215 (1978-07-01)
Eight fluorescent dye combinations for simultaneous DNA-protein staining have been evaluated spectroscopically and flow microfluorometrically: propidium iodide (PI) with fluoresceinisothiocyanate (FITC), fluorescamine (FC), and dansylchloride (DANS); diamidinophenylindole (DAPII) with sulphorhodamin (SR101), tetramethylrhodamin isothiocyanate (TRITC), and nitrobenzodiazole (NBD); acriflavine (AF) with
Zheqing Cai et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17462-17467 (2013-10-09)
Both preclinical and clinical studies suggest that brief cycles of ischemia and reperfusion in the arm or leg may protect the heart against injury following prolonged coronary artery occlusion and reperfusion, a phenomenon known as remote ischemic preconditioning. Recent studies
Nadine Fischer et al.
Biochimica et biophysica acta, 1828(2), 632-641 (2012-10-24)
Acriflavine resistance protein B acts as the active transporter in the multi-drug efflux pump Acriflavine resistance proteins A / B - Tolerance to colicins protein in Escherichia coli. Within the same reaction cycle intermediate all Acriflavine resistance protein B X-ray



Global Trade Item Number

SKUGTIN
PH000036-25MG04061831294828
01673-10G04061838609571
01673-50G04061838609588

Questions

Reviews

No rating value

Active Filters