Skip to Content
Merck
All Photos(4)

Key Documents

A8251

Sigma-Aldrich

Acriflavine hydrochloride

Powder

Synonym(s):

3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
6265525
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Acriflavine hydrochloride, fluorescent dye

Quality Level

form

powder

composition

Acriflavine HCL content, 10-50%
Proflavine HCL, 25-70%

technique(s)

microbe id | staining: suitable

color

faint brown to brown, and Orange to Dark Orange and Red to Very Dark Red

mp

260 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cl[H].Cl[H].Nc1ccc2cc3ccc(N)cc3nc2c1.C[n+]4c5cc(N)ccc5cc6ccc(N)cc46

InChI

1S/C14H13N3.C13H11N3.3ClH/c1-17-13-7-11(15)4-2-9(13)6-10-3-5-12(16)8-14(10)17;14-10-3-1-8-5-9-2-4-11(15)7-13(9)16-12(8)6-10;;;/h2-8H,1H3,(H3,15,16);1-7H,14-15H2;3*1H

InChI key

MKLTXAHQKDVBLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

In living mice, acriflavine hydrochloride has been used in the confocal laser endomicroscopy to check the crypt structure in mucosa. It has also been used in Neurospora strain selection by checking the acriflavine resistance.

Biochem/physiol Actions

Acriflavine hydrochloride is a fluorescent dye. It is used as a topical contrast agent. It mainly stains nuclei and is applied to tissues before microscopy.

Other Notes

A mixture of 3,6-diamino-10-methylacridinium (acriflavin) and 3,6-diaminoacridine (proflavin)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

period-1 encodes an ATP-dependent RNA helicase that influences nutritional compensation of the Neurospora circadian clock.
Emerson JM, et al.
Proceedings of the National Academy of Sciences of the USA, 112, 15707-15712 (2015)
Jian Yang et al.
Journal of the American Heart Association, 7(5) (2018-02-27)
Limb remote ischemic preconditioning (RIPC) protects against brain injury induced by stroke, but the underlying protective mechanisms remain unknown. As hypoxia inducible factor 1α (HIF-1α) is neuroprotective in stroke and mediates neuroinflammation, we tested the hypothesis that HIF-1α is a
Confocal laser endomicroscopy and narrow-band imaging-aided endoscopy for in vivo imaging of colitis and colon cancer in mice.
Waldner MJ, et al.
Nature Protocols, 6, 1471-1481 (2011)
Nashwa Amin et al.
Cells, 10(6) (2021-07-03)
Hypoxia-inducible factor 1 can sufficiently control the progress of neurological symptoms after ischemic stroke owing to their actions associated with its downstream genes. In this study, we evaluated the role of HIF-1α in attenuating brain damage after endothelin-1 injection. Focal
Confocal endomicroscopy: in vivo diagnosis of neoplastic lesions of the gastrointestinal tract.
Goetz M and Kiesslich R
Anticancer Research, 28, 353-360 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service