Skip to Content
Merck

Skip To

UST157

Aliphatics Mix (C5-C12)

certified reference material, 2000 μg/mL each component in methanol

Synonym(s):

C5-C12 Aliphatic Hydrocarbon Mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC UST-157

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UST127UST122UST155
application(s)

environmental
petroleum

application(s)

environmental
petroleum

application(s)

environmental

application(s)

environmental
petroleum

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

Quality Level

100, 300

Quality Level

100, 300

Quality Level

100, 300

Quality Level

100, 300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

concentration

2000 μg/mL each component in methanol

concentration

2000 μg/mL in dichloromethane: carbon disulfide (50:50)

concentration

1000 μg/mL in methylene chloride

concentration

5000 μg/mL in methanol


Analyte

Description

  • Pentane

  • Hexane

  • Heptane

  • Octane

  • Nonane

  • Decane

  • Undecane

  • Dodecane

See All (8)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F - closed cup - Solvent

flash_point_c

11 °C - closed cup - Solvent



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
UST157-1.5ML04061837618734

Questions

Reviews

No rating value

Active Filters