Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of × 1.5 mL
manufacturer/tradename
RTC UST-122
concentration
1000 μg/mL in methylene chloride
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-30°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Analyte
Description
- Acenaphthene
- Acenaphthylene
- Anthracene
- Benz[a]anthracene
- Benzo[b]fluoranthene
- Benzo[ghi]perylene
- Benzo[k]fluoranthene
- Chrysene
- Dibenz[a,h]anthracene
- Fluoranthene
- Fluorene
- Indeno[1,2,3-cd]pyrene
- Naphthalene
- Phenanthrene
- Pyrene
See All (15)
signalword
Danger
hcodes
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


