Skip to Content
Merck

Skip To

D6750

Sodium deoxycholate

≥97% (titration)

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H39NaO4
CAS Number:
Molecular Weight:
414.55
UNSPSC Code:
12161900
NACRES:
NA.21
PubChem Substance ID:
EC Number:
206-132-7
Beilstein/REAXYS Number:
3581950
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

assay

≥97% (titration)

form

powder

mol wt

micellar avg mol wt 1200-5000

aggregation number

3-12

CMC

2-6 mM (20-25°C)

solubility

water: 50 mg/mL, clear, colorless to light yellow

HLB

16

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC([O-])=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1

InChI key

FHHPUSMSKHSNKW-SMOYURAASA-M

General description

Deoxycholic acid sodium salt, also known as Sodium Deoxycholate, is a highly versatile ionic detergent utilized for its exceptional ability to solubilize membrane-bound proteins and lyse cells. Its mechanism involves disrupting cell membranes and dissolving hydrophobic molecules, making it an invaluable reagent in scientific research applications. In protein research, it plays a pivotal role in solubilizing proteins, particularly those embedded in membranes, enabling their extraction, purification, and subsequent analysis.

Moreover, Sodium Deoxycholate extends its utility to nucleic acid extraction from cells and tissues. By lysing cells and disrupting cellular structures, it facilitates the release and purification of nucleic acids, proving valuable in cell biology and biochemical research. Beyond these applications, its effectiveness in cell lysis processes positions it as a valuable tool for breaking down cells and releasing their contents, finding applications in diverse biological studies, encompassing microscopy, cell biology, and biochemical research.

Application

Sodium deoxycholate has been used in the lysis buffer prepared for brown adipose tissue.[1] It has been used in the decellularization of aortic valvular conduit.[2]

Biochem/physiol Actions

Solubilizes fats for absorption in the intestines.

Features and Benefits

Highly versatile surfactant for your cell biology and biochemical research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
30970264103264101
form

powder

form

powder

form

solid

form

powder

assay

≥97% (titration)

assay

≥98.0% (dry matter, NT)

assay

≥99% (TLC)

assay

≥96% (TLC)

aggregation number

3-12

aggregation number

3-12

aggregation number

3 - 12, 3-12

aggregation number

3-12, 4 - 10

storage temp.

room temp

storage temp.

-

storage temp.

15-25°C

storage temp.

15-25°C

mol wt

micellar avg mol wt 1200-5000

mol wt

micellar avg mol wt 1200-5000

mol wt

micellar avg mol wt 1200-5000, micellar wt 1200-4900

mol wt

micellar avg mol wt 1200-5000, micellar wt 1700-4200

HLB

16

HLB

16

HLB

16

HLB

16


Still not finding the right product?

Explore all of our products under Sodium deoxycholate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How safe is this product when used on animals?

    1 answer
    1. This product is intended for research use only and is not suitable for human or veterinary use. While not tested for in vitro or in vivo use, this product has been used successfully in animal models by other customers. Please see the links below to review example publications:

      Corroboration of naringin effects on the intestinal absorption and pharmacokinetic behavior of candesartan cilexetil solid dispersions using in-situ rat models.
      https://pubmed.ncbi.nlm.nih.gov/24918161/

      MicroRNA-101 inhibits rat cardiac hypertrophy by targeting Rab1a.
      https://pubmed.ncbi.nlm.nih.gov/25850725/

      A model of metabolic syndrome and related diseases with intestinal endotoxemia in rats fed a high fat and high sucrose diet.
      https://pubmed.ncbi.nlm.nih.gov/25502558/

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters