Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium deoxycholate
Synonym(s):
3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt, Sodium deoxycholate
Empirical Formula (Hill Notation):
C
24
H
39
NaO
4
CAS Number:
302-95-4
Molecular Weight:
414.55
EC Number:
206-132-7
Products
Product Category
detergents (3)
inhibitors (1)
Brand
Millipore (2)
SAFC (1)
Sigma-Aldrich (3)
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Tissue
Quality Segment
Functional Group
Application
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
30970
Sodium deoxycholate,
BioXtra, ≥98.0% (dry matter, NT)
Expand
View Pricing
S1827
Sodium deoxycholate,
Expand
View Pricing
264103
Sodium deoxycholate,
ULTROL
®
Grade, ≥99% (TLC), detergent to solubilize membrane-bound proteins in their native state
Expand
View Pricing
D6750
Sodium deoxycholate,
≥97% (titration)
Expand
View Pricing
1.06504
Sodium deoxycholate,
Bile salt and ionic detergent. Supplement. For the isolation and classification of gram-negative bacteria
Expand
View Pricing
SRE0046
Sodium deoxycholate,
Suitable for manufacturing of diagnostic kits and reagents
Expand
View Pricing