Skip to Content
Merck

Skip To

46211

2-Pentanone

analytical standard

Synonym(s):

Methyl propyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3COCH2CH2CH3
CAS Number:
Molecular Weight:
86.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-528-1
Beilstein/REAXYS Number:
506058
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor pressure

27 mmHg ( 20 °C)

assay

98-100% (GC)

autoignition temp.

941 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.2 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.39 (lit.)

bp

101-105 °C (lit.)

mp

−78 °C (lit.)

solubility

H2O: soluble 72.6 g/L at 20 °C (OECD Test Guideline 105)

density

0.809 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCC(C)=O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W284220471194W284203
application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

food and beverages

application(s)

flavors and fragrances

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable

technique(s)

-

format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

300, 400

grade

analytical standard

grade

FG, Fragrance grade, Halal, Kosher, natural

grade

HPLC grade

grade

FG, Halal

assay

98-100% (GC)

assay

≥99.0%

assay

99.5%

assay

≥98%


Still not finding the right product?

Explore all of our products under 2-Pentanone


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

46.4 °F - closed cup

flash_point_c

8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J M Mathews et al.
Toxicology and applied pharmacology, 146(2), 255-260 (1997-11-05)
The effect of trans-1,2-dichloroethylene (DCE), an inhibitor of cytochrome P450 (P450) 2E1 (CYP2E1), on the composition and quantity of volatile organic chemicals (VOCs) expired in the breath of male F-344 rats was determined in parallel with hepatic P450 activity and
Jenny Pettersson et al.
Biological & pharmaceutical bulletin, 31(3), 534-537 (2008-03-04)
Many phytochemicals found in the diet may prevent colon carcinogenesis by affecting biochemical processes in the colonic mucosa. Inflammation and subsequent elevation of the enzyme cyclooxygenase-2 (COX-2) are two such factors involved in the development of colon cancer, and inhibition
Linus Olofsson et al.
Organic & biomolecular chemistry, 3(5), 750-755 (2005-02-26)
The enantioselective reduction of 2-pentanone to (R)- and (S)-2-pentanol by Thermoanaerobacter (formerly Thermoanaerobium) brockii alcohol dehydrogenase (TBADH) in mixtures of water and water-miscible organic solvents was investigated. Significant enzymatic activity was retained in up to 87% methanol, ethanol and acetonitrile.



Global Trade Item Number

SKUGTIN
46211-5ML04061832346366

Questions

Reviews

No rating value

Active Filters