Skip to Content
Merck
All Photos(3)

Key Documents

471194

Sigma-Aldrich

2-Pentanone

suitable for HPLC, 99.5%

Synonym(s):

Methyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
506058
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
101-105 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)

grade

HPLC grade

vapor pressure

27 mmHg ( 20 °C)

Assay

99.5%

form

liquid

autoignition temp.

941 °F

expl. lim.

8.2 %

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.0003%

refractive index

n20/D 1.39 (lit.)

bp

101-105 °C (lit.)

mp

−78 °C (lit.)

solubility

water: 72.6 g/L at 20 °C (OECD Test Guideline 105)

density

0.809 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 330 nm Amax: 1.00
λ: 340 nm Amax: 0.10
λ: 350 nm Amax: 0.01
λ: 370-400 nm Amax: 0.005

application(s)

food and beverages

SMILES string

CCCC(C)=O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fadda et al.
FEMS microbiology letters, 210(2), 209-214 (2002-06-05)
Staphylococcus carnosus strain 833, inoculated into sausage, increased the levels of methyl ketones which contributed to the cured aroma. These ketones were predicted to arise from incomplete beta-oxidation followed by a decarboxylation. To check this hypothesis, we measured the beta-decarboxylase
Yosuke Hanai et al.
Bioscience, biotechnology, and biochemistry, 76(4), 679-684 (2012-04-10)
Lung cancer is a leading cause of deaths in cancer. Hence, developing early-stage diagnostic tests that are non-invasive, highly sensitive, and specific is crucial. In this study, we investigated to determine whether biomarkers derived from urinary volatile organic compounds (VOCs)
Silvina G Fadda et al.
Journal of agricultural and food chemistry, 50(9), 2471-2474 (2002-04-18)
Methyl ketones are detected in dry fermented sausages in which they contribute to the cured aroma. They have been associated with the inoculation of Staphylococcus carnosus used as starter culture. To evaluate the ability of bacterial starters to produce methyl
Yasuhisa Asano et al.
Bioscience, biotechnology, and biochemistry, 69(12), 2349-2357 (2005-12-27)
We established a simple HPLC method to determine the activity and stereochemistry of the chiral mandelonitrile synthesized from benzaldehyde and cyanide, and applied it to screen for hydroxynitrile lyase (HNL) activity of plant origin. A total of 163 species of
J M Mathews et al.
Toxicology and applied pharmacology, 146(2), 255-260 (1997-11-05)
The effect of trans-1,2-dichloroethylene (DCE), an inhibitor of cytochrome P450 (P450) 2E1 (CYP2E1), on the composition and quantity of volatile organic chemicals (VOCs) expired in the breath of male F-344 rats was determined in parallel with hepatic P450 activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service