Skip to Content
Merck

Skip To

44504

Dowex 50W X8 Ion Exchange Resin

hydrogen form, 16-50 mesh, strongly acidic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
NB.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dowex 50W X8, hydrogen form, strongly acidic, 16-50 mesh

description

cross-linkage 8%

Quality Level

form

beads

moisture

50-58%

technique(s)

LPLC: suitable

matrix

styrene divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

16-50 mesh

capacity

1.1 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

also called HCR-W

Application

Dowex 50WX8 (HCR-W) hydrogen form is used:
  • As stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted.
  • As a catalyst.
  • In deionizing supernatant fraction samples for paper chromatography.

Dowex 50WX8 hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.

Legal Information


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
445192174921.05221
matrix

styrene divinylbenzene (gel)

matrix

styrene divinylbenzene (gel)

matrix

styrene-divinylbenzene (gel)

matrix

PS-DVB

separation technique

cation exchange

separation technique

cation exchange

separation technique

cation exchange

separation technique

cation exchange

matrix active group

sulfonic acid functional group

matrix active group

sulfonic acid functional group

matrix active group

sulfonic acid functional group

matrix active group

SCX

particle size

16-50 mesh

particle size

200-400 mesh

particle size

50-100 mesh

particle size

90-250 μm

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

HPLC: suitable, LPLC: suitable

form

beads

form

beads

form

beads

form

resin


Still not finding the right product?

Explore all of our products under Dowex 50W X8


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Novel ion exchange chromatographic method using conductimetric detection.
Small, Hamish, Timothy S. Stevens, and William C. Bauman.
Analytical Chemistry, 47, 1801-1809 (1975)
Chemical composition and ultrastructure of wild-type and white mutantAspergillus nidulans conidial walls.
Claverie-Martin, Felix, Maria R. Diaz-Torres, and Michael J. Geoghegan.
Current Microbiology, 16, 281-287 (1988)
Hydrogen generation from hydrolysis of sodium borohydride using Ni-Ru nanocomposite as catalysts.
Liu, Cheng-Hong, et al.
International Journal of Hydrogen Energy, 34, 2153-2163 (2009)



Global Trade Item Number

SKUGTIN
44504-100G04061838514707

Questions

1–10 of 15 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. Is this resin capable of separating choline

    1 answer
    1. No references are available for using the hydrogen form of this resin to separate choline from other relevant cations. However, ion-exchange chromatography is suitable for this separation. Reviewing peer-reviewed literature on this topic is recommended. Contact Technical Service for assistance with detailed method development.

      Helpful?

  3. Is this type of resin suitable for separating ammonia gas from air?

    1 answer
    1. This product would not be recommended for separation of ammonia gas from air. Typically, a scrubbing system with a chemical such as dilute sulfuric acid would be used. If an adsorbent is preferred, activated charcoal would be the best recommendation and impregnated activated charcoal is ideal.

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  5. What is the particle size and shape of Product 44504?

    1 answer
    1. The mesh size (20-50 mesh) corresponds to a particle size range of approximately 300 to 850 microns. The product is in the form of beads.

      Helpful?

  6. What information do you have on the stability of Dowex® 50W X8 resin, Product 44504?

    1 answer
    1. The ion exchange resin is tolerant of a wide range of chemical conditions, including most inorganic and organic solutions with exception of strong oxidizing agents. Solutions, such as nitric or chromic acid, bleach or chlorine-generating solutions as well as peroxide should be avoided as these will result in rapid oxidation. Exposure of resins to metal ions, such as iron, manganese and copper should be minimized in oxidizing environments as they can catalyze the oxidation.

      Helpful?

  7. What is the matrix for the Dowex® 50W X8 resin, Product 44504?

    1 answer
    1. The fine mesh resin is based on a microporous copolymer of styrene and divinylbenzene (DVB) (styrene-DVB gel).

      Helpful?

  8. What are the proper storage conditions for Dowex® 50W X8 resin, Product 44504?

    1 answer
    1. The resins should be stored in a dry cool place. The storage temperature should be kept below 50oC (120oF).Storage temperature below 0oC can cause resin freezing and temperatures below -18oC (0oF) should be avoided. Repeated freeze-thaw can cause damage on the beads. The frozen resin beads should be completely thawed out at room temperature condition before loading and use.

      Helpful?

  9. What is the suggested operating temperature for Dowex® 50W X8 resin, Product 44504?

    1 answer
    1. At elevated temperature, operation can cause degradation of functional sites and loss of the ion exchange capacity. The resin degradation  depends on many variables, such as  the level of dissolved oxygen and the presence of catalytic metals on the resin. In general, ~120oC (250oF) is suggested as the maximum operating temperature.

      Helpful?

  10. What is the form of Product 44504, Dowex® 50W X8?

    1 answer
    1. This resin is a strongly acidic cation exchanger, in the H+ ionic form.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters