Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dowex 50W-X8 Ion Exchange Resin
CAS Number:
69011-20-7
Products
Product Category
resins (3)
columns (1)
Brand
Supelco (2)
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.05221
Dowex 50W-X8 Ion Exchange Resin,
hydrogen form, 60-170 mesh, strongly acidic, cationic
Expand
View Pricing
44504
Dowex 50W-X8 Ion Exchange Resin,
hydrogen form, 16-50 mesh, strongly acidic
Expand
View Pricing
44519
Dowex 50W-X8 Ion Exchange Resin,
hydrogen form, 200-400 mesh, strongly acidic
Expand
View Pricing
44509
Dowex 50W-X8 Ion Exchange Resin,
hydrogen form, 50-100 mesh, strongly acidic
Expand
View Pricing