Skip to Content
Merck

Skip To

08992

3,4-Dihydroxybenzoic acid

analytical standard

Synonym(s):

Protocatechuic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
202-760-0
Beilstein/REAXYS Number:
1448841
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

197-200 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)c1ccc(O)c(O)c1

InChI

1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)

InChI key

YQUVCSBJEUQKSH-UHFFFAOYSA-N

General description

3,4-Dihydroxybenzoic acid is a catabolite of epinephrine. It is a phenolic compound and an important constituent of lignin present in wine, green and black tea, and herbal medicines, which exhibits anti-inflammatory and antitumor promotion. 3,4-Dihydroxybenzoic acid is also a model matrix for ionization of peptides, proteins and carbohydrates in the MALDI (matrix-assisted laser desorption ionization mass spectroscopy) technique.

Application

3,4-Dihydroxybenzoic acid may be used as a reference standard for the determination of:
  • Phenols in biomass pre-treatment hydrolysates by high-performance liquid chromatography method.
  • 3,4-Dihydroxybenzoic acid in wine by high-performance liquid chromatography with a microbore column.

Biochem/physiol Actions

Chemopreventive in several animal models of carcinogenesis. Blocks cell proliferation in the post-initiation phase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
37580W44309503930590
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

analytical standard

grade

-

grade

FG

grade

primary reference standard

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

flavors and fragrances

application(s)

food and beverages

format

neat

format

-

format

-

format

-

assay

≥97.0% (HPLC)

assay

≥97.0% (T)

assay

≥99%

assay

-

mp

197-200 °C (dec.) (lit.)

mp

197-200 °C (dec.) (lit.)

mp

197-200 °C (dec.) (lit.)

mp

197-200 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under 3,4-Dihydroxybenzoic acid


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Thermochemical study of 2, 4-, 2, 6-and 3, 4-dihydroxybenzoic acids in the liquid phase using a TG apparatus.
Vecchio S and Brunetti B
Thermochimica Acta, 515(1), 84-90 (2011)
Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
3, 4-Dihydroxybenzoic acid from Smilacis chinae rhizome protects amyloid $\beta$ protein (25-35)-induced neurotoxicity in cultured rat cortical neurons.
Ban JY, et al.
Neuroscience Letters, 420(2), 184-188 (2007)



Global Trade Item Number

SKUGTIN
286508-1G04061826264737
08992-50MG04061838653024

Questions

Reviews

No rating value

Active Filters