Skip to Content
Merck
All Photos(1)

Key Documents

37580

Sigma-Aldrich

3,4-Dihydroxybenzoic acid

≥97.0% (T)

Synonym(s):

Protocatechuic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1448841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0% (T)

mp

197-200 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc(O)c(O)c1

InChI

1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)

InChI key

YQUVCSBJEUQKSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3,4-Dihydroxybenzoic acid is reported as dietery polyphenol.[1] Antiviral activity of 3,4-dihydroxybenzoic acid (protocatechuic acid, PCA) against a virulent H9N2 strain in a mouse model is reported.[2] Cytotoxic effects of PCA on 3 non-small cell lung cancer (NSCLC) cell lines, A549, H3255, and Calu-6 cell lines is reported.[3] PCA is reported to prevent the carcinogenesis or antitumor growth in vivo. Signaling pathway involved in PCA-induced apoptosis in human gastric adenocarcinoma (AGS) cells is reported.[4] The oxidation of 3,4-dihydroxybenzoic acid by H2O2 in aqueous/goethite slurry at varying operating conditions (catalyst load, temperature, pH, substrate and hydrogen peroxide starting concentration) is investigated.[5]

Application

3,4-Dihydroxybenzoic acid is suitable for use in a study to investigate the effect of 4-coumaric and 3,4-dihydroxybenzoic acid on the basal oxidative DNA damage of rat colonic mucosa in vivo.[1] It may be used in the derivatization of chitosan resin.[6]

Biochem/physiol Actions

Chemopreventive in several animal models of carcinogenesis. Blocks cell proliferation in the post-initiation phase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Andreozzi et al.
Water research, 36(11), 2761-2768 (2002-07-31)
The oxidation by H2O2 of 3,4-dihydroxybenzoic acid (3,4-DHB) in aqueous/goethite slurry at varying operating conditions (catalyst load, temperature, pH, substrate and hydrogen peroxide starting concentration) is investigated. At adopted catalyst loads the observed kinetic developments are consistent with a non-radicalic
Hui-Hsuan Lin et al.
International journal of cancer, 120(11), 2306-2316 (2007-02-17)
3,4-Dihydroxybenzoic acid (protocatechuic acid, PCA) is discussed to represent antioxidative food components in a human diet rich in fruits and vegetables, and has been shown to prevent carcinogenesis or antitumor growth in vivo. However, the molecular mechanisms involved in chemopreventive
Akhmad Sabarudin et al.
Analytica chimica acta, 581(2), 214-220 (2007-03-28)
A chitosan resin derivatized with 3,4-dihydroxybenzoic acid moiety (CCTS-DHBA resin) was newly synthesized for the collection/concentration of trace uranium by using cross-linked chitosan (CCTS) as base material, and the adsorption behavior of uranium as well as 60 elements on the
Shih-ming Tsao et al.
Nutrition and cancer, 66(8), 1331-1341 (2014-10-31)
Cytotoxic effects of protocatechuic acid (PCA) upon 3 nonsmall cell lung cancer (NSCLC) cell lines, A549, H3255, and Calu-6 cell lines, were examined. PCA at 1, 2, 4, and 8 μM was used to treat these cells. Results showed that
Francesco Guglielmi et al.
The British journal of nutrition, 89(5), 581-587 (2003-05-02)
The effect of 4-coumaric and 3,4-dihydroxybenzoic (protocatechuic) acid on the basal oxidative DNA damage of rat colonic mucosa in vivo was studied, relative to vitamin E. F344 rats were treated with 4-coumaric or protocatechuic acid mixed in the diet (25

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service