Yes, the solvent for this product is water.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Product Name
Sodium azide 0.1 M solution,
Quality Segment
density
1.00 g/mL at 20 °C
storage temp.
2-8°C
SMILES string
[Na]N=[N+]=[N-]
InChI
1S/N3.Na/c1-3-2;/q-1;+1
InChI key
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Application
Sodium azide 0.1M solution has been used as an inhibitor of ceruloplasmin activity.
Biochem/physiol Actions
Sodium azide is an F-type ATPase inhibitor and selectively inhibits ceruloplasmin.
Sodium azide 0.1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
Sodium azide 0.1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| density 1.00 g/mL at 20 °C | density 1.85 g/cm3 at 20 °C | density - | density - |
| storage temp. 2-8°C | storage temp. 15-25°C | storage temp. - | storage temp. - |
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Clinical Environmental Health and Toxic Exposures (2001)
Active immunization against vasoactive intestinal polypeptide decreases neuronal recruitment and inhibits reproduction in zebra finches.
Vistoropsky Y
The Journal of Comparative Neurology, 524, 2516-2516 (2016)
Ceruloplasmin dysfunction and therapeutic potential for Parkinson disease.
Ayton S et al.
Annals of Neurology, 73, 554-554 (2013)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 261548-250ML | 04061837886409 |
| 261548-1L | 04061837886393 |
| 759406-10ML | 04061837889592 |
| 759406-50ML | 04061837889608 |
| 08591-1ML-F | 04061838652621 |
-
What is the solvent? Water?
1 answer-
Helpful?
-
-
What is the percentage of the 0.1 M Sodium azide?
1 answer-
Sodium Azide has a molecular weight of 65.01 g/mol. A 1M solution would be a 6.5% solution. A 0.1M solution would be a 0.65% solution
Helpful?
-
-
what's the product volume of 08591, Sodium azide 0.1 M solution? 1 mL/each?
1 answer-
The package size of this item is 1mL.
Helpful?
-
