Skip to Content
Merck
All Photos(1)

Key Documents

1.06688

Sigma-Aldrich

Sodium azide

extra pure, Technipur®

Synonym(s):

Sodium azide, Hydrazoic azid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
N3Na
CAS Number:
Molecular Weight:
65.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
247-852-1
NACRES:
NA.21

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

potency

27 mg/kg LD50, oral (Rat)

bp

300 °C/1013 hPa (rigorous decomposition)

mp

275 °C (decomposition)

solubility

420 g/L

density

1.85 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Na+].[N+](=[N-])=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium azide (NaN3) is an inorganic azide. It is widely used as an active ingredient in automobile airbag inflators. It is also used in detonators, explosives and as a preservative in biological solutions.

Application

Sodium azide can be used as:
  • A component of a preservative named azidiol (a mixture of chloramphenicol and NaN3), which is used in preserving raw milk in the dairy industry.
  • A biocide to prevent deterioration of cane juice in sugar industries.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deterioration of cane juice?sources and indicators
Eggleston G
Food Chemistry, 78(1), 95-103 (2002)
Electrochemical behaviour of Ti?6Al?4V alloy and Ti in azide and halide solutions
Author links open overlay panel
Heakal F E-T, et al.
Corrosion Science, 53(9), 2728-2737 (2011)
Environmental Fate of Sodium Azide Derived from Automobile Airbags
Betterton EA
Critical reviews in environmental science and technology, 33(4), 423-458 (2003)
Interactions of sodium azide with triazine herbicides: Effect on sorption to soils
Author links open overlay panel
Chefetz B
Chemosphere, 65(2), 352-357 (2006)
Optimization of Preservation Methods Allows Deeper Insights into Changes of Raw Milk Microbiota
Ouamba AJK, et al.
Microorganisms, 8(3), 368-368 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service