Skip to Content
Merck

Skip To

05112

Activated charcoal

suitable for GC

Synonym(s):

Charcoal activated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
NACRES:
SB.54
PubChem Substance ID:
UNSPSC Code:
23201100
EC Number:
231-153-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

form

granular

autoignition temp.

842 °F

resistivity

1375 μΩ-cm, 20°C (graphite)

technique(s)

LPLC: suitable, gas chromatography (GC): suitable

matrix

Carbon

matrix active group

carbon

particle size

0.3-0.5 mm

mp

3550 °C (lit.)

bulk density

0.4‑0.5 g/cm3

separation technique

reversed phase

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

Activated charcoal or activated carbon is nontoxic in nature and is used as an adsorbent to remove broad spectrum of pollutants in air, water and soil. It does not lose potency when kept in container. It has no odour or taste. It can be prepared from any carboneous material using chemical or gas activation methods. Gas activation is preferred when the raw material has less than 25% moisture content.

Application

It may be used for fiber coating multiwalled carbon nanotubes to compare its solid-phase microextraction efficiency in water and milk samples before GC-ECD analysis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C288980485161551
form

granular

form

untreated, granular

form

powder

form

powder

particle size

0.3-0.5 mm

particle size

8-20 mesh

particle size

50-150 μm

particle size

-100 (mesh)

matrix

Carbon

matrix

-

matrix

Carbon

matrix

-

mp

3550 °C (lit.)

mp

3550 °C (lit.)

mp

3550 °C (lit.)

mp

3550 °C (lit.)

technique(s)

LPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

LPLC: suitable

technique(s)

-

vapor pressure

<0.1 mmHg ( 20 °C)

vapor pressure

<0.1 mmHg ( 20 °C)

vapor pressure

<0.1 mmHg ( 20 °C)

vapor pressure

<0.1 mmHg ( 20 °C)


Still not finding the right product?

Explore all of our products under Activated charcoal


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Activated charcoal; preparation, characterization and applications: a review article.
Mohammad-Khah, A., and R. Ansari.
International Journal of ChemTech Research, 1, 2745-2788 (2009)
David O. Cooney
Activated Charcoal: Antidote, Remedy and Health Aid, 102-102 (2016)
Multiwalled carbon nanotubes coated fibers for solid-phase microextraction of polybrominated diphenyl ethers in water and milk samples before gas chromatography with electron-capture detection.
Wang, Jun-Xia, et al
Journal of Chromatography A, 1137.1, 8-14 (2006)



Global Trade Item Number

SKUGTIN
810953000004022536403564
810953000104022536403571
810953999904022536403588
295809-1G04061826649022
810953000504022536622422
05112-500G04061833438619
05112-100G04061833378984

Questions

  1. What is the typical pore volume for this activated carbon?

    1 answer
    1. Unfortunately, the pore volume is not determined. The surface area by BET is 900 m²/g.

      Helpful?

Reviews

No rating value

Active Filters