Skip to Content
Merck
All Photos(1)

Key Documents

80485

Supelco

Activated Charcoal Carbon Adsorbent

for the determination of AOX, 100-285 mesh

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.54

product name

Activated charcoal, for the determination of AOX, 50-150 μm particle size

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

form

powder

autoignition temp.

842 °F

resistivity

1375 μΩ-cm, 20°C (graphite)

technique(s)

LPLC: suitable

loss

≤20% loss on drying

matrix

Carbon

matrix active group

carbon

particle size

50-150 μm

mp

3550 °C (lit.)

application(s)

environmental

separation technique

reversed phase

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal is a black powder that is generally obtained by subjecting wood or other carbonaceous materials to pyrolysis process.

Application

Activated charcoal may be used as an analytical standard for the determination of adsorbable organic halides (AOX) in:
  • Landfill leachates, glyphosate formulations, and treated wastewater by adsorption-pyrolysis-microcoulometric procedure.
  • Aqueous environmental samples by adsorption followed by combustion ion chromatography (CIC).
  • Surface water and wastewater samples by adsorption-pyrolysis followed by plasma emission spectrometric detection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated charcoal revisited.
Gaudreault P.
Clinical Pediatric Emergency Medicine, 6(2) (2005)
Electrochemical degradation of glyphosate formulations at [TM="DSA"] anodes in chloride medium: an AOX formation study
Neto SA and De Andrade AR
J. Appl. Electrochem., 39(10), 1863-1863 (2009)
Quality of treated wastewater: method validation of AOX.
Cherif S, et al.
Accreditation and Quality Assurance, 11(12), 632-637 (2006)
Development of an element-selective monitoring system for adsorbable organic halogens (AOX) with plasma emission spectrometric detection for quasi-continuous waste-water analysis.
Twiehaus T, et al.
Fresenius Journal of Analytical Chemistry, 371(5), 614-620 (2001)
Determination of adsorbable organic fluorine from aqueous environmental samples by adsorption to polystyrene-divinylbenzene based activated carbon and combustion ion chromatography.
Wagner A, et al.
Journal of Chromatography A, 1295(12), 82-89 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service