Skip to Content
Merck

Skip To

TRYPSEQ-RO

Roche

Trypsin Sequencing Grade

from bovine pancreas

Synonym(s):

trypsin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
≥80 units/mg protein
Biological source:
bovine pancreas
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas

form

lyophilized (salt-free)

specific activity

≥80 units/mg protein

mol wt

Mr 23.5 kDa

packaging

pkg of 4 × 100 μg (11047841001), pkg of 4 × 25 μg (11418475001)

manufacturer/tradename

Roche

optimum pH

8.0

storage temp.

2-8°C

General description

Trypsin Sequencing Grade is isolated from bovine pancreas as a highly purified and specific protease. Trypsin Sequencing Grade is a serine endopeptidase.[1] It specifically cleaves peptide bonds at the carboxylic side of the basic amino acids Arg and Lys. Amide and ester bonds of Arg and Lys are also cleaved.[2]

Application

Trypsin Sequencing Grade is used to to digest proteins in solution, in gels or on blotting membranes.
The enzyme is used for protein-structure elucidation, tryptic mapping, fingerprinting, sequence analysis, and translocation studies.[3] Trypsin Sequencing Grade generates glycopeptides from purified glycoproteins and is suited for the digestion of proteins in polyacrylamide gels.

Biochem/physiol Actions

The specificity of Trypsin Sequencing Grade is verified with the oxidized B-chain of insulin (insulin Box) as substrate. High concentrations of Trypsin Sequencing Grade (1 part by weight enzyme with 18 parts by weight insulin Box) are incubated for 18 hours to detect traces of chymotrypsin impurities.

Specificity (HPLC, with Insulin Box):
Cleavage after 1 hour: ≥90%;
Unspecific cleavage products after 18 hours: ≤10%

Preparation Note

Stabilizers: Trypsin is stable in 4 M.
Working concentration: 1/100 to 1/20 of the protein by weight (in solution); 1-5 μg/100 μl (for in-gel digest)
Storage conditions (working solution): A solution in 0.01% trifluoroacetic acid (TFA), (v/v) or 1 mM HCl may be used for one week at maximum, if stored at 2 to 8 °C. By incubation of proteins in solution at neutral to slightly basic pH-values partial autolysis might occur. For this application, Roche recommends Trypsin, Modified, Sequencing Grade.
Reconstitution in acid is necessary for stability of solution: 0.01% TFA (v/v), 1 mM HCl or 0.1% acetic acid are recommended.

Analysis Note

Purity: Free of impurities that may interfere with the separation of peptides in reversed-phase HPLC.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TRYPSEQM-RORTRYP-RO650275
specific activity

≥80 units/mg protein

specific activity

-

specific activity

≥150 units/mg protein

specific activity

≥2.5 units/mg protein

biological source

bovine pancreas

biological source

bovine pancreas

biological source

-

biological source

-

form

lyophilized (salt-free)

form

lyophilized (salt-free)

form

lyophilized

form

lyophilized solid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

Mr 23.5 kDa

mol wt

24.000 g/mol

mol wt

-

mol wt

-

optimum pH

8.0

optimum pH

8.0

optimum pH

-

optimum pH

-


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Saeedreza Vessal et al.
Journal of proteome research, 11(8), 4289-4307 (2012-07-07)
Protein expression patterns in imbibed seeds of three cultivars of chickpea (Cicer arietinum L.) with different rates of germination under limiting water supply in soil (>10% water holding capacity) were compared. A large number of soluble proteins expressed earlier and
Petra A Neff et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 8(14), 2133-2137 (2007-08-28)
A silicon-on-insulator (SOI) based thin film resistor is employed for the label-free determination of enzymatic activity. We demonstrate that enzymes, which cleave biological polyelectrolyte substrates, can be detected by the sensor. As an application, we consider the serine endopeptidase trypsin
Jesper V Olsen et al.
Molecular & cellular proteomics : MCP, 3(6), 608-614 (2004-03-23)
Almost all large-scale projects in mass spectrometry-based proteomics use trypsin to convert protein mixtures into more readily analyzable peptide populations. When searching peptide fragmentation spectra against sequence databases, potentially matching peptide sequences can be required to conform to tryptic specificity



Global Trade Item Number

SKUGTIN
1141847500104061838250841
1104784100104061838261373