Skip to Content
Merck
All Photos(1)

Key Documents

650275

Sigma-Aldrich

Trypsin, Iodination Grade, Human Pancreas

Synonym(s):

Trypsin, Iodination Grade, Human Pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.21
Pricing and availability is not currently available.

description

Merck USA index - 14, 9795

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized solid

specific activity

≥2.5 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

HCl: 2 mM
aqueous buffer, pH 2.5: soluble

foreign activity

Chymotrypsin, none detected

shipped in

wet ice

storage temp.

−20°C

General description

Native trypsin from human pancreas. Serine protease that specifically hydrolyzes amides, peptides, and proteins at the C-terminus side of arginine and lysine residues.

Warning

Toxicity: Irritant (B)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of N-benzyl-L-arginine-pNA per min at 25°C, 200 mM Tris-HCl, pH 7.8, 20 mM CaCl₂.

Physical form

Salt-free.

Preparation Note

Prepared from tissue that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C).

Other Notes

Merck Index13, 9865.
Travis, J., and Roberts, R.C. 1969. Biochemistry 8, 2884.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

when assayed against N-Suc-Ala-Ala-Pro-Phe-pNA (573462)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service